Skip to Content
Merck
All Photos(2)

Key Documents

5.38254

Sigma-Aldrich

GN44028

Synonym(s):

GN44028, N -(2,3-Dihydrobenzo[b][1,4]dioxin-6-yl)-1,4-dihydroindeno[1,2-c]pyrazol-3-amine, HIF-1 Inhibitor II

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H15N3O2
Molecular Weight:
305.33
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Assay

≥98% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

brown

solubility

DMSO: 50 mg/mL

storage temp.

2-8°C

General description

A cell-permeable indenopyrazole based compound that potently suppresses hypoxia-induced HIF-1α transcription (IC50 = 14 nM in HRE-Luc HeLa cells) and VEGF-expression. Shown to effectively induce growth arrest in several cancer cells (IC50 = 2.1, 3.7 & 1.8 µM for HCT116, HepG2 & HeLa cells), although with reduced efficiency in PC3 (IC50 = 25.4 µM). Does neither affect HIF-1α protein accumulation nor HIF-1α/HIF-1β heterodimer complex formation. Suggested to act via impairing HIF-1α/HIF-1β heterodimer-induced transcriptional pathway.

Please note that the molecular weight for this compound is batch-specific due to variable water content. Please refer to the vial label or the certificate of analysis for the batch-specific molecular weight. The molecular weight provided represents the baseline molecular weight without water.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
HIF-1α
Reversible: yes
Secondary Target
VEGF
Target IC50: 14 nM in HRE-Luc HeLa cells

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Minegishi, H., et al. 2013. ACS Med. Chem. Lett.4, 297.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service