Skip to Content
Merck
All Photos(2)

Key Documents

5.33060

Sigma-Aldrich

HMGB Inhibitor, Inflachromene

Synonym(s):

HMGB Inhibitor, Inflachromene, Neuroinflammatory Inhibitor, Inflachromene, HMGB2 Inhibitor, Inflachromene, 10-Hydroxy-7,7-dimethyl-2-phenyl-7,12b-dihydrochromeno[4,3-c][1,2,4]triazolo[1,2-a]pyridazine-1,3(2H,5H)-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H19N3O4
Molecular Weight:
377.39
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥98% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 50 mg/mL

storage temp.

−20°C

SMILES string

CC1(C2=CCN3C(=O)N(C(=O)N3C2C4=C(O1)C=C(C=C4)O)C5=CC=CC=C5)C

General description

A cell-permeable, chromene-fused benzopyran-embedded tetracyclic compound that selectively and directly binds to the DNA binding domain box A of high mobility group box (HMGB) proteins and blocks their post-translational phosphorylation and acetylation. Diminishes the cytoplasmic accumulation of HMGB2 in microglial cells. Blocks lipopolysaccharide (LPS)-induced nitrite release in BV-2 microglial cells, RAW 264.7 macrophages, and mouse primary microglial cultures (~50 nM to 20 µM) without inducing any toxicity. However, it is shown to be less potent in primary astrocytes. Suppresses LPS-stimulated increase in the levels of IL-6, IL-1b, NOS2, and TNFα genes and inhibits the nuclear translocation of NF-κB. Also shown to reduce LPS-induced phosphorylation of ERK, JNK, and p38 MAP kinases in microglia. Reduces neuroinflammation, neurite damage, and spinal cord demyelination in a murine model of auto-immune encephalitis (~10 mg/kg).

Please note that the molecular weight for this compound is batch-specific due to variable water content. Please refer to the vial label or the certificate of analysis for the batch-specific molecular weight. The molecular weight provided represents the baseline molecular weight without water.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
HMGB2
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Lee, S., et al. 2014. Nat. Chem. Biol.10, in press.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service