Skip to Content
Merck
All Photos(1)

Documents

5.31492

Sigma-Aldrich

12/15-Lipoxygenase Inhibitor, ML351

Synonym(s):

12/15-Lipoxygenase Inhibitor, ML351, 5-(Methylamino)-2-(naphthalen-1-yl)oxazole-4-carbonitrile, 12/15-LOX Inhibitor, 15-Lipoxygenase-1 Inhibitor

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H11N3O
CAS Number:
Molecular Weight:
249.27
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥97% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 100 mg/mL

storage temp.

2-8°C

General description

A cell-permeable, non-toxic naphthyl-oxazole compound that acts as a highly potent, selective, active site directed, mixed type inhibitor of lipoxygenase-12/15 (IC50 = 200 nM for human). The inhibition appears to be non-reductive and reversible in nature. Displays excellent selectivity over 5-LOX (IC50 >50 µM), 12-LOX (IC50 >100 µM), and 15-LOX (IC50 >100 µM) and does not affect the activity of COX-1 and -2 in any significant manner (<10% inhibition at 15 µM). Can cross the blood brain barrier (brain to plasma ratio = 2.8). Offers protection against glutamate-induced oxidative toxicity in mouse neuronal HT22 cells and reduces the size of infarcts in a murine model of ischemic stroke. Displays good aqueous stability over a wide range of pH. Exhibits attractive pharmacokinetic properties (t1/2 ~ 1 h in both plasma and brain; Cmax of 13.8 and 28.8 µM in plasma and brain, respectively; at 30 mg/kg, i.p).
A cell-permeable, non-toxic naphthyl-oxazole compound that acts as a highly potent, selective, active site directed, mixed type inhibitor of lipoxygenase-12/15 (IC50 = 200 nM for human). The inhibition appears to be non-reductive and reversible in nature. Displays excellent selectivity over 5-LOX (IC50 >50 µM), 12-LOX (IC50 >100 µM), and 15-LOX (IC50 >100 µM) and does not affect the activity of COX-1 and -2 in any significant manner (<10% inhibition at 15 µM). Can cross the blood brain barrier (brain to plasma ratio = 2.8). Offers protection against glutamate-induced oxidative toxicity in mouse neuronal HT22 cells and reduces the size of infarcts in a murine model of ischemic stroke. Displays good aqueous stability over a wide range of pH. Exhibits attractive pharmacokinetic properties (t1/2 ~ 1 h in both plasma and brain; Cmax of 13.8 and 28.8 µM in plasma and brain, respectively; at 30 mg/kg, i.p).

Please note that the molecular weight for this compound is batch-specific due to variable water content.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
12/15-LOX
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Rai, G., et al. 2014. J. Med. Chem.57, 4035.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service