Skip to Content
Merck
All Photos(1)

Documents

5.31145

Sigma-Aldrich

mGlu7 Antagonist, XAP044

Synonym(s):

mGlu7 Antagonist, XAP044, 7-Hydroxy-3-(4-iodophenoxy)-4H-chromen-4-one, 7-Hydroxy-3-(4-iodophenoxy)-chromen-4-one, Metabotropic Glutamate Receptor Subtype 7 Antagonist, XAP-044

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H9IO4
CAS Number:
Molecular Weight:
380.13
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥97% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

pale yellow

solubility

DMSO: 100 mg/mL

storage temp.

2-8°C

InChI

1S/C15H9IO4/c16-9-1-4-11(5-2-9)20-14-8-19-13-7-10(17)3-6-12(13)15(14)18/h1-8,17H

InChI key

XXZKJJYWINSUMJ-UHFFFAOYSA-N

General description

A blood-brain barrier permeable and bioavailable chromenone compound that acts as a potent, selective, and reversible antagonist of mGlu7 receptor (IC50 = 3.5 and 2.8 µM for mGlu7a and mGlu7b in CHO cells). Docks within the Venus flytrap domain (VFTD) close to the binding site of L-glutamate and functions as an orthosteric-like antagonist. Reduces DL-2-amino-4-phosphonobutyric acid (DL-AP4)-induced activation of human mGlu7b receptor (IC50 = 2.8 µM), but does not affect DL-AP4-induced activation of [35S]GTPγS binding by mGlu4 and mGlu6. Shown to diminish [35S]GTPγS binding by the human mGlu7/6 chimeric receptor harboring mGlu7 VFTD (IC50 = 2.5 µM). Displays weak binding affinity towards mGlu5 and mGlu8 receptors (IC50 >20 and 33 µM, respectively) and has no effect on GABA-induced activation of [35S]GTPγS binding by GABA-B receptors. Displays antidepressant and anxiolytic-like properties and selectively blocks long-term potentiation (LTP) in the lateral amygdala of wild type (IC50 = 88 nM), but not of mGlu7- deficient mice.
A blood-brain barrier permeable and bioavailable chromenone compound that acts as a potent, selective, and reversible antagonist of mGlu7 receptor (IC50 = 3.5 and 2.8 µM for mGlu7a and mGlu7b in CHO cells). Docks within the Venus flytrap domain (VFTD) close to the binding site of L-glutamate and functions as an orthosteric-like antagonist. Reduces DL-2-amino-4-phosphonobutyric acid (DL-AP4)-induced activation of human mGlu7b receptor (IC50 = 2.8 µM), but does not affect DL-AP4-induced activation of [35S]GTPγS binding by mGlu4 and mGlu6. Shown to diminish [35S]GTPγS binding by the human mGlu7/6 chimeric receptor harboring mGlu7 VFTD (IC50 = 2.5 µM). Displays weak binding affinity towards mGlu5 and mGlu8 receptors (IC50 >20 and 33 µM, respectively) and has no effect on GABA-induced activation of [35S]GTPγS binding by GABA-B receptors. Displays antidepressant and anxiolytic-like properties and selectively blocks long-term potentiation (LTP) in the lateral amygdala of wild type (IC50 = 88 nM), but not of mGlu7- deficient mice.

Please note that the molecular weight for this compound is batch-specific due to variable water content.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
mGlu7R
Reversible: yes
Target IC50: 3.5 and 2.8 &micro

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Gee, C.E., et al. 2014. J. Biol. Chem.289, 10975.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service