Skip to Content
Merck
All Photos(1)

Key Documents

5.31088

Sigma-Aldrich

VCP Inhibitor III, NMS-873

Synonym(s):

VCP Inhibitor III, NMS-873, 3-(3-(Cyclopentylthio)-5-((2-methyl-4ʹ-(methylsulfonyl)biphenyl-4-yloxy)methyl)-4H-1,2,4-triazol-4-yl)pyridine, 3-(3-Cyclopentylsulfanyl-5-(4ʹ-methanesulfonyl-2-methyl-biphenyl-4-yloxymethyl)-[1,2,4]triazol-4-yl)-pyridine, Valosin-containing Protein Inhibitor III, p97 Inhibitor III, ERAD Inhibitor III, NMS873

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H28N4O3S2
CAS Number:
Molecular Weight:
520.67
MDL number:
UNSPSC Code:
12352200
Pricing and availability is not currently available.

Assay

≥98% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

light beige

storage temp.

2-8°C

InChI

1S/C27H28N4O3S2/c1-19-16-22(11-14-25(19)20-9-12-24(13-10-20)36(2,32)33)34-18-26-29-30-27(35-23-7-3-4-8-23)31(26)21-6-5-15-28-17-21/h5-6,9-17,23H,3-4,7-8,18H2,1-2H3

InChI key

UJGTUKMAJVCBIS-UHFFFAOYSA-N

General description

A cell-permeable triazolylpyridine compound that potently inhibits VCP ATPase activity in a non-ATP-competitive manner (IC50/[ATP] = 20 nM/1 mM & 30 nM/60 µM; [wt hVCP] = 10 nM) via an allosteric mechanism where the inhibitor targets the lateral tunnel formed by the D1 & D2 domains of one VCP protomer together with the D1 from the adjacent VCP molecule in the hexamer, effectively locking the protomer in a rigid conformation and thereby preventing the release of bound ADP for successful propagation of interprotomer ATP hydrolysis cycle, while being ineffective against a panel of 53 kinases or the ATPase activities of 4 other AAA (ATPase Associated with diverse/various cellular Activities) ATPases (NSF, SPATAS, VPS4B, and cdc6) and Hsp90 (by <15% at 10 µM). In agreement with siRNA-mediated VCP knockdown, NMS-873 is shown to prevent VCP from extracting ubiquitinated client proteins, including cyclin E & Mcl-1, for proteasomal degradation, resulting in cellular UPR (unfold protein response) activation, autophagy induction, and eventual apoptosis in HCT-116 cultures in a dose- and time-dependent manner (0.5 to 5 µM; 8 to 72 h). Anti-cancer studies using a panel of 40 various hematological and solid tumor lines reveal wide-spread sensitivities to NMS-873-induced killings (IC50 from 80 nM to 2 µM), differing in degrees than those observed with the proteasome inhibitor Bortezomib (Cat. No. Cat. No. 504314). Exhibits high clearance and low bioavailability in in vivo studies.

Please note that the molecular weight for this compound is batch-specific due to variable water content.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
VCP
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Magnaghi, P., et al. 2013. Nat. Chem. Biol.9, 548.
Polucci, P., et al. 2013. J. Med. Chem.56, 437.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service