Skip to Content
Merck
All Photos(1)

Key Documents

5.30588

Sigma-Aldrich

IκB Ubiquitination Inhibitor, GS143

Synonym(s):

IκB Ubiquitination Inhibitor, GS143, 4-(3-Benzyl-4-(5-(2-fluoro-phenyl)-furan-2-ylmethylene)-5-oxo-4,5-dihydropyrazol-1-yl)-benzoic acid, 4-(3-Benzyl-4-((5-(2-fluorophenyl)furan-2-yl)methylene)-5-oxo-4,5-dihydro-1H-pyrazol-1-yl)benzoic acid, GS 143

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H19FN2O4
CAS Number:
Molecular Weight:
466.46
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

Assay

≥97% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

dark red

solubility

DMSO: 25 mg/mL

storage temp.

2-8°C

SMILES string

C1=CC=C(C=C1)CC2=NN(C(=O)C2=CC3=CC=C(O3)C4=CC=CC=C4F)C5=CC=C(C=C5)C(=O)O

General description

A cell-permeable trisubstituted oxo-dihydropyrazolyl-benzoic acid that selectively inhibits the E3 ligase complex SCFβTrCP1-mediated ubiquitination of phosphorylated IκBα (IC50 = 5.2 µM), displaying little inhibitory potency against IKKβ-catalyzed IκBα phosphorylation, MDM2-mediated p53 ubiquitination, or the protease activities of cathepsin B, chymotrypsin, 20S & 26S proteasome (IC50 >100 µM). Cellular IκBα degradation blockage by GS143 treatment (complete inhibition against TNFα-induced degradation with 30 min 20 µM drug pretreatment in in HeLa S3 and HT29 cultures) effectively inhibits TNFα- and LPS-stimulated NF-κB transcription in HEK293, HT-29, and THP-1 cultures (IC50 from 2.1 to 10.5 µM). Intranasal administration (32 µg/20 µL/mouse) among OVA-sensitized mice 2 h prior to OVA challenge via inhalation greatly suppress OVA challenge-induced NF-κB activation in lung tissue (>90% inhibition of nuclear NF-κB p65 DNA-binding activity 2 h post challenge) and airway inflammation (>80% reduction in eosinophils & lymphocytes in BALF 48 h post challenge) in vivo. GS143 most likely prevents SCFβTrCP1 from interacting with phosphorylated IκBα without inhibiting SCFβTrCP1 E3 ligase activity directly, as no apparent accumulation of another known SCFβTrCP1 substrate β-catenin is seen in HeLa S3 cells upon GS143 treatment (up to 20 µM for 3 h).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
phosphorylated IκBα

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.
Use only fresh DMSO for reconstitution.

Other Notes

Hirose, K., et al. 2008. Biochem. Biophys. Res. Commun.374, 507.

Nakajima, H., et al. 2008. Biochem. Biophys. Res. Commun.368, 1007.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service