Skip to Content
Merck
All Photos(1)

Key Documents

528282

Sigma-Aldrich

Polo-like Kinase Inhibitor I

The Polo-like Kinase Inhibitor I, also referenced under CAS 916985-21-2, controls the biological activity of Polo-like Kinase. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

Polo-like Kinase Inhibitor I, 5-(5,6-Dimethoxybenzimidazol-1-yl)-3-(4-methanesulfonyl-benzyloxy)-thiophene-2-carboxamide, Plk Inhibitor I, Plk Inhibitor I, 5-(5,6-Dimethoxybenzimidazol-1-yl)-3-(4-methanesulfonyl-benzyloxy)-thiophene-2-carboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H21N3O6S2
CAS Number:
Molecular Weight:
487.55
UNSPSC Code:
12352200

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

tan

solubility

DMSO: 10 mg/mL

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C22H21N3O6S2/c1-29-17-8-15-16(9-18(17)30-2)25(12-24-15)20-10-19(21(32-20)22(23)26)31-11-13-4-6-14(7-5-13)33(3,27)28/h4-10,12H,11H2,1-3H3,(H2,23,26)

InChI key

YYZTVTZCYVZGRB-UHFFFAOYSA-N

General description

A thiophenecarboxamide compound that acts as a potent and ATP-binding site-targeting inhibitor of polo-like kinase 1 (IC50 = 126 nM) with excellent selectivity over IKK-2 and IKK-3 (IC50 >16 µM).

Biochem/physiol Actions

Cell permeable: no
Primary Target
Polo-like Kinase
Product does not compete with ATP.
Reversible: no
Target IC50: 126 nM against polo-like kinase 1

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Bamborough, P., et al. 2006. Bioorg. Med. Chem. Lett.16, 6236.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service