Skip to Content
Merck
All Photos(1)

Key Documents

528244

Millipore

Platensimycin, Streptomyces sp.

A cell-permeable Streptomyces-derived antibiotic that exhibits broad-spectrum Gram-positive antibacterial activity by selectively targeting the elongation condensing enzyme FabF.

Synonym(s):

Platensimycin, Streptomyces sp.

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H27NO7
CAS Number:
Molecular Weight:
441.47
UNSPSC Code:
12352200

Quality Level

Assay

≥93% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

tan

solubility

DMSO: 20 mg/mL
ethanol: 20 mg/mL

shipped in

wet ice

storage temp.

−20°C

InChI

1S/C24H27NO7/c1-22(7-6-17(28)25-18-14(26)4-3-13(19(18)29)21(30)31)16(27)5-8-24-10-12-9-15(20(22)24)32-23(12,2)11-24/h3-5,8,12,15,20,26,29H,6-7,9-11H2,1-2H3,(H,25,28)(H,30,31)/t12-,15+,20+,22-,23+,24+/m1/s1

General description

A cell-permeable Streptomyces-derived antibiotic that exhibits broad-spectrum Gram-positive antibacterial activity by selectively targeting the elongation condensing enzyme FabF (IC50 = 48 and 160 nM against S. aureus and E. coli FabF, respectively), but not the initiation condensing enzyme FabH (IC50 = 67 µM), involved in type II fatty acid synthesis (FASII). Binding studies indicate that acyl-FabF intermediate complex formation induces a FabF conformation change that is necessary for Platensimycin interaction. Platensimycin effectively kills numerous Staphylococcus aureus, Enterococcus faecium, and Streptococcus pneumoniae strains, including the ones that are resistant to methicillin and vancomycin (Cat. No. 627850). Platensimycin does not exhibit antifungal activity towards Candida albicans (no effect at 64 µg/ml) or toxicity toward mammalian HeLa culture (no effect at 1000 µg/ml) and is efficacious in treating S. aureus infection in mice in vivo (~105-fold bacteria titre reduction via a 24 h i.v. at 150 µg h-1). Although Platensimycin is ineffective toward wild-type E. coli due to the presence of functional multidrug efflux pump (no effect at 64 µg/ml), Platensimycin does inhibit the growth of efflux-negative E. coli strains.
A cell-permeable Streptomyces-derived antibiotic that exhibits broad-spectrum Gram-positive antibacterial activity by selectively targeting the elongation condensing enzyme FabF (IC50 = 48 and 160 nM against S. aureus and E. coli FabF, respectively), but not the initiation condensing enzyme FabH (IC50 = 67 µM), involved in type II fatty acid synthesis (FASII). Binding studies indicate that acyl-FabF intermediate complex formation induces a FabF conformation change that is necessary for Platensimycin interaction. Platensimycin effectively kills numerous Staphylococcus aureus, Enterococcus faecium, and Streptococcus pneumoniae strains, including the ones that are resistant to methicillin and vancomycin (Cat. No. 627850). Platensimycin does not exhibit antifungal activity towards Candida albicans (no effect at 64 µg/ml) or toxicity toward mammalian HeLa culture (no effect at 1000 µg/ml) and is efficacious in treating S. aureus infection in mice in vivo (~105-fold bacteria titre reduction via a 24 h i.v. at 150 µg h-1). Although Platensimycin is ineffective toward wild-type E. coli due to the presence of functional multidrug efflux pump (no effect at 64 µg/ml), Platensimycin does inhibit the growth of efflux-negative E. coli strains. Platensimycin has been shown to potently and selectively inhibit hepatocyte FAS and fatty acid oxidation (FAO), without affecting sterol synthesis. Platensimycin is also known as Fatty Acid Synthase Inhibitor III.

Warning

Toxicity: Harmful (C)

Other Notes

Wu, M., et al. 2011. Proc. Natl. Acad. Sci. USAin press.
Wang, J., et al. 2007. Proc. Natl. Acad. Sci. USA104, 7612.
Singh, S.B., et al. 2006. J. Am. Chem. Soc.128, 11916.
Wang, J., et al. 2006. Nature441, 358.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service