Skip to Content
Merck
All Photos(1)

Key Documents

528150

Sigma-Aldrich

PARP Inhibitor VIII, PJ34

The PARP Inhibitor VIII, PJ34, also referenced under CAS 344458-15-7, controls the biological activity of PARP. This small molecule/inhibitor is primarily used for Cell Structure applications.

Synonym(s):

PARP Inhibitor VIII, PJ34

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H17N3O2 · xHCl
CAS Number:
Molecular Weight:
295.34 (free base basis)
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

white

solubility

water: 5 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

Cl.[nH]1c2c(c3c([c]1=O)cccc3)cc(cc2)NC(=O)CN(C)C

InChI

1S/C17H17N3O2.ClH/c1-20(2)10-16(21)18-11-7-8-15-14(9-11)12-5-3-4-6-13(12)17(22)19-15;/h3-9H,10H2,1-2H3,(H,18,21)(H,19,22);1H

InChI key

RURAZZMDMNRXMI-UHFFFAOYSA-N

General description

A cell-permeable, water-soluble phenanthridinone-derivative that acts as a potent inhibitor of poly(ADP-ribose) polymerase (PARP; EC50 = 20 nM). Shown to be about 10,000 times more potent than the prototypical PARP inhibitor, 3-Aminobenzamide (Cat. No. 165350 ; EC50 = 200 µM). Does not act as an antioxidant at higher concentrations (1 µM to 10 mM). Exhibits neuroprotection in in vivo and in vitro models of stroke. Also available as a 20 mM solution in H2O(Cat. No. 528151 ).

Biochem/physiol Actions

Cell permeable: yes
EC50 = 20 nM inhibiting poly(ADP-ribose) polymerase (PARP)
Primary Target
PARP
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Abdelkarim, G.E., et al. 2001. Int. J. Mol. Med.7, 255.
Garcia-Soriano, F.G., et al. 2001. Nature Med. 7, 108.
Mabley, G.J., et al. 2001. Inflamm. Res.50, 561.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joelle C Boulos et al.
Acta pharmacologica Sinica, 44(11), 2265-2281 (2023-06-22)
The majority of blood malignancies is incurable and has unforeseeable remitting-relapsing paths in response to different treatments. Cynaropicrin, a natural sesquiterpene lactone from the edible parts of the artichoke plant, has gained increased attention as a chemotherapeutic agent. In this
Pablo Iglesias et al.
International journal of molecular sciences, 24(10) (2023-05-27)
In recent years, new therapies have been developed based on molecules that target molecular mechanisms involved in both the initiation and maintenance of the oncogenic process. Among these molecules are the poly(ADP-ribose) polymerase 1 (PARP1) inhibitors. PARP1 has emerged as
Han-Wei Liu et al.
Neuron, 111(10), 1609-1625 (2023-03-16)
Stroke prognosis is negatively associated with an elevation of serum bilirubin, but how bilirubin worsens outcomes remains mysterious. We report that post-, but not pre-, stroke bilirubin levels among inpatients scale with infarct volume. In mouse models, bilirubin increases neuronal
Shiori Kuraoka et al.
Molecular & cellular proteomics : MCP, 100153-100153 (2021-10-01)
Mass spectrometry-enabled ADP-ribosylation workflows are developing rapidly, providing researchers a variety of ADP-ribosylome enrichment strategies and mass spectrometric acquisition options. Despite the growth spurt in upstream technologies, systematic ADP-ribosyl (ADPr) peptide mass spectral annotation methods are lacking. HCD-dependent ADP-ribosylome studies

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service