Skip to Content
Merck
All Photos(1)

Documents

524617

Sigma-Aldrich

L-α-Lecithin

≥98% (TLC), liquid, lipotropic agent, Calbiochem

Synonym(s):

L-α-Lecithin, Egg Yolk, Highly Purified, L-α-Phosphatidylcholine

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.77

product name

L-α-Lecithin, Egg Yolk, Highly Purified, Chromatographically purified.

Quality Level

Assay

≥98% (TLC)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

solubility

chloroform: soluble

shipped in

ambient

storage temp.

−20°C

General description

Chromatographically purified. A lipotropic agent.

Biochem/physiol Actions

Cell permeable: no
Primary Target
A lipotropic agent
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas
Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Flammable (J)

Physical form

In Chloroform:MeOH (3:2), 0.1% BHT at 100 mg/ml.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Other Notes

Due to the nature of the Hazardous Materials in this shipment, additional shipping charges may be applied to your order. Certain sizes may be exempt from the additional hazardous materials shipping charges. Please contact your local sales office for more information regarding these charges.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 1 - STOT SE 3

Target Organs

Eyes,Central nervous system, Liver,Kidney, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

53.6 °F

Flash Point(C)

12 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Irina S R Rio et al.
Pharmaceutics, 13(11) (2021-11-28)
Liposome-like nanoarchitectures containing manganese ferrite nanoparticles covered or decorated with gold were developed for application in dual cancer therapy, combining chemotherapy and photothermia. The magnetic/plasmonic nanoparticles were characterized using XRD, UV/Visible absorption, HR-TEM, and SQUID, exhibiting superparamagnetic behavior at room

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service