Skip to Content
Merck
All Photos(1)

Key Documents

506153

Sigma-Aldrich

p38 MAP Kinase Inhibitor IV

The p38 MAP Kinase Inhibitor IV, also referenced under CAS 1638-41-1, controls the biological activity of p38 MAP Kinase. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

p38 MAP Kinase Inhibitor IV, 2,2ʹ-Sulfonyl-bis-(3,4,6-trichlorophenol), MT4

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H4Cl6O4S
CAS Number:
Molecular Weight:
456.94
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

light yellow

solubility

DMSO: 4.5 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

[S](=O)(=O)(c2c(c(cc(c2O)Cl)Cl)Cl)c1c(c(cc(c1O)Cl)Cl)Cl

InChI

1S/C12H4Cl6O4S/c13-3-1-5(15)9(19)11(7(3)17)23(21,22)12-8(18)4(14)2-6(16)10(12)20/h1-2,19-20H

InChI key

AKUKHICVNCCQHN-UHFFFAOYSA-N

General description

A cell-permeable symmetrical sulfone compound that acts as a potent and ATP-competitive inhibitor of p38α/β MAPK (IC50 = 130 and 550 nM, respectively), while exihibiting much reduced activity (≤23% inhibition at 1 µM) against p38γ/δ, ERK1/2, and JNK1/2/3. Shown to be more effective than SB 203850 (Cat. Nos. 559389, 559395, and 559398) in inhibiting LPS-induced IL-1β release from hPBMC (100% vs. 50% inhibition with 100 µM respective inhibitor).
A cell-permeable symmetrical sulfone compound that acts as a potent and ATP-competitive inhibitor of p38α/β MAPK (IC50 = 130 and 550 nM, respectively), while exihibiting much reduced activity (≤23% inhibition at 1 µM) against p38γ/δ, ERK1/2, and JNK1/2/3. Shown to be more effective than SB 203850 (Cat. Nos. 559389, 559395, and 559398) in inhibiting LPS-induced IL-1β release from hPBMC (100% vs. 50% inhibition with 100 µM respective inhibitor).

Packaging

Packaged under inert gas

Warning

Toxicity: Harmful (C)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Liu, Y.C., et al. 2007. Biochem. Biophys. Res. Commun.352, 656.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Patricia Fernández-Nogueira et al.
Cancer letters, 424, 70-83 (2018-03-20)
Histamine receptor 1 (HRH1) belongs to the rhodopsin-like G-protein-coupled receptor family. Its activation by histamine triggers cell proliferation, embryonic development, and tumor growth. We recently established that HRH1 is up-regulated in basal and human epidermal growth factor receptor 2 (HER2)-enriched

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service