Skip to Content
Merck
All Photos(1)

Documents

505487

Sigma-Aldrich

TEPP-46

≥98% (HPLC), solid, PKM2 activator, Calbiochem®

Synonym(s):

PKM2 Activator IV, TEPP-46, 6-((3-Aminophenyl)methyl)-4-methyl-2-methylsulfinylthieno[3,4]pyrrolo[1,3-d]pyridazin-5-one, Pyruvate Kinase M2 Activator IV, ML265, NCGC00186528, TEPP46, Pyruvate Kinase M2 Activator IV, ML265, NCGC00186528, TEPP46, 6-((3-Aminophenyl)methyl)-4-methyl-2-methylsulfinylthieno[3,4]pyrrolo[1,3-d]pyridazin-5-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H16N4O2S2
CAS Number:
Molecular Weight:
372.46
UNSPSC Code:
12352200
NACRES:
NA.77

product name

PKM2 Activator IV, TEPP-46,

Assay

≥98% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 100 mg/mL

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C17H16N4O2S2/c1-20-13-7-14(25(2)23)24-16(13)12-8-19-21(17(22)15(12)20)9-10-4-3-5-11(18)6-10/h3-8H,9,18H2,1-2H3

InChI key

ZWKJWVSEDISQIS-UHFFFAOYSA-N

General description

A cell-permeable thienopyrrolopyridazinone that acts as a potent allosteric PKM2 activator (AC50 = 92 nM; [PKM2] = 0.1 nM) via stoichiometric interaction (Two activator molecules per tetramer) with FBP-bound PKM2 tetramer (One FBP per monomer) in a similar manner as PKM2 Activator III (Cat. No. 504537), resulting in highly stabilized PKM2 tetramer resistant to dissociation in the presence of phosphotyrosine-containing peptide/protein or upon PKM2 K305Q mutation. Reported to exhibit good aqueous solubility (29.6 µg/mL or 79.5 µM in PBS, pH 7.4) and retard human lung carcinoma H1299 proliferation under hypoxia, but not normoxia, condition in cultures (doubling time = 50 and 35 h, respectively, with or without 30 µM TEPP-46; 1% O2) in vitro. TEPP-46 is bioavailable in mice via i.p. (Tmax = 0.5 h; Cmax = 10.854 µg/mL; AUCINF = 26.994 hr·µg/mL; 10 mg/kg) or p.o. (Tmax = 0.25 h; Cmax = 10.179 µg/mL; AUCINF = 24.350 hr·µg/mL; 10 mg/kg) administration and shown to impair H1299 tumorigenesis in mice (%tumor development/av. tumor weight = 30%/0.13 mg and 70%/2.6 mg, respectively, with or without 50 mg/kg/12 h oral dosage for 7 wks) in vivo with no apparent toxicity.
A cell-permeable thienopyrrolopyridazinone that exhibits good aqueous solubility (29.6 µg/mL or 79.5 µM in PBS, pH 7.4) and acts as a potent allosteric PKM2 activator (AC50 = 92 nM; [PKM2] = 0.1 nM) by stabilizing PKM2 tetramer via stoichiometric interaction with FBP-bound PKM2 (2 activator/4 FBP/tetramer) in a similar manner as PKM2 Activator III (Cat. No. 504537). Reprorted to retard human lung carcinoma H1299 proliferation under hypoxia, but not normoxia, condition in cultures (doubling time = 50 and 35 h, respectively, with or without 30 µM TEPP-46; 1% O2) in vitro and impair H1299 tumorigenesis in mice (50 mg/kg/12 h p.o.) in vivo with no apparent toxicity.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
PKM2
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.
Use only fresh DMSO for reconstitution.

Other Notes

Anastasiou, D., et al. 2012. Nat. Chem. Biol.8, 839.

Jiang, J.K., et al. 2010. Bioorg. Med. Chem. Lett.20, 3387.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Beth A Griesel et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 35(7), e21728-e21728 (2021-06-11)
Proliferation and differentiation of preadipocytes, and other cell types, is accompanied by an increase in glucose uptake. Previous work showed that a pulse of high glucose was required during the first 3 days of differentiation in vitro, but was not required
Samuel R Taylor et al.
Nature, 597(7875), 263-267 (2021-08-20)
Fructose consumption is linked to the rising incidence of obesity and cancer, which are two of the leading causes of morbidity and mortality globally1,2. Dietary fructose metabolism begins at the epithelium of the small intestine, where fructose is transported by
Flávio P Veras et al.
Cell reports, 41(13), 111897-111897 (2022-12-29)
Psoriasis is an inflammatory skin disease characterized by keratinocyte proliferation and inflammatory cell infiltration induced by IL-17. However, the molecular mechanism through which IL-17 signaling in keratinocytes triggers skin inflammation remains not fully understood. Pyruvate kinase M2 (PKM2), a glycolytic
Sheyda Bahiraii et al.
Frontiers in immunology, 13, 935692-935692 (2022-08-20)
Murine macrophages activated by the Toll-like receptor 4 agonist lipopolysaccharide (LPS) polarize to the M1 type by inducing proinflammatory marker proteins and changing their energy metabolism to increased aerobic glycolysis and reduced respiration. We here show that the aliphatic isothiocyanate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service