Skip to Content
Merck
All Photos(1)

Key Documents

5.05413

Sigma-Aldrich

5-Iodo-A-85380 dihydrochloride hydrate

Synonym(s):

5-Iodo-A-85380 dihydrochloride hydrate, 3-((2S)-2-Azetidinylmethoxy)-5-iodo-pyridine dihydrochloride hydrate, α4β2 and α6β2 Nicotinic Acetylcholine Receptors (nAChR) Agonist, 5-Iodo-A-85380

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H11IN2O · 2HCl · xH2O
CAS Number:
Molecular Weight:
363.02 (anhydrous basis)
UNSPSC Code:
12352200

Assay

≥98% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

water: 22 mg/mL

storage temp.

2-8°C

InChI

1S/C9H11IN2O/c10-7-3-9(5-11-4-7)13-6-8-1-2-12-8/h3-5,8,12H,1-2,6H2/t8-/m0/s1

InChI key

RKVRGRXOYDTUEY-QMMMGPOBSA-N

General description

A very potent agonist highly selective for α4β2 and α6β2 nicotinic acetylcholine receptors (nAChR) (Ki= 12 and 10 pM for human and rat α4β2 nAChRs, respectively). An important nAChR ligand for the treatment of pain, smoking addition and other mental disorders.

Biochem/physiol Actions

Primary Target
α4β2 nAChR

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Liu, X., et al., 2013. Psychopharm.In press.

Kennett, A., et al., 2012. Nicotine Tob. Res.14, 1339.

Huang. LZ, et al., 2011. Neuropharm.60, 861.
Mogg. AJ, et al., 2004. Neuropharm.60, 861.

Mukhin, A.G., et al., 2000. Mol. Pharmacol.47, 848.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service