Skip to Content
Merck
All Photos(1)

Documents

5.05378

Sigma-Aldrich

IKK Inhibitor VII

InSolution, ≥97%, 10 mM in DMSO, potent, selective, and ATP-competitive inhibitor of IKK

Synonym(s):

InSolution IKK Inhibitor VII, (4-(4-(Benzo[b]thiophen-2-yl)pyrimidin-2-ylamino)phenyl)(4-(pyrrolidin-1-yl)piperidin-1-yl)methanone, InSolution IKK Inhibitor VII, IKK 16, IKK 16, (4-(4-(Benzo[b]thiophen-2-yl)pyrimidin-2-ylamino)phenyl)(4-(pyrrolidin-1-yl)piperidin-1-yl)methanone, InSolution IKK Inhibitor VII

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H29N5OS
CAS Number:
Molecular Weight:
483.63
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥97% (HPLC)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles
desiccated (hygroscopic)
protect from light

storage temp.

−70°C

InChI

1S/C28H29N5OS/c34-27(33-17-12-23(13-18-33)32-15-3-4-16-32)20-7-9-22(10-8-20)30-28-29-14-11-24(31-28)26-19-21-5-1-2-6-25(21)35-26/h1-2,5-11,14,19,23H,3-4,12-13,15-18H2,(H,29,30,31)

InChI key

BWZJBXAPRCVCKQ-UHFFFAOYSA-N

General description

A cell-permeable benzamido-pyrimidine compound that acts as a potent, selective, and ATP-competitive inhibitor of IKK (IC50 = 40, 70, and 200 nM for IKK-2, IKK complex, and IKK-1, respectively). Inhibits cellular IκBα degradation and NF-κB-mediated gene expression in vitro in HUVEC cells and has been shown to exhibit excellent in vivo efficacy both in mice and rats.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
IKK-2, IKK complex, and IKK-1
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Irritant (B)

Physical form

A 10 mM (500 µg/103 µL) sterile-filtered solution of IKK Inhibitor VII (Cat. No. 401486) in DMSO.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C). Aliquots are stable for up to 6 months at -20°C.

Other Notes

Waelchli, R., et al. 2006. Bioorg. Med. Chem. Lett.16, 108.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service