Skip to Content
Merck
All Photos(1)

Key Documents

5.04913

Sigma-Aldrich

Aniracetam

Synonym(s):

Aniracetam, 1-(4-methoxybenzoyl)pyrrolidin-2-one, Draganon, Sarpul, Ampamet, Memodrin, Referan, Memodrin, Referan, 1-(4-methoxybenzoyl)pyrrolidin-2-one, Draganon, Sarpul, Ampamet

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H13NO3
CAS Number:
Molecular Weight:
219.24
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Assay

≥99% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 100 mM

storage temp.

2-8°C

SMILES string

N2(CCCC2=O)C(=O)c1ccc(cc1)OC

InChI

1S/C12H13NO3/c1-16-10-6-4-9(5-7-10)12(15)13-8-2-3-11(13)14/h4-7H,2-3,8H2,1H3

InChI key

ZXNRTKGTQJPIJK-UHFFFAOYSA-N

General description

A positive allosteric modulator selective for AMPA receptors. Slows down AMPA receptor deactivation rate without altering the agonist potency. A leading candidate of ampakine nootropic for cognitive enhancement. Widely used in studies in a variety of brain functions (i.e. learn and memory, cognition) and disorders (depression, anxiety disorder, Alzheimer′s disease, and aging).

Please note that the molecular weight for this compound is batch-specific due to variable water content.

Biochem/physiol Actions

Primary Target
AMPA receptors

Warning

Toxicity: Standard Handling (A)

Other Notes

Koliaki, C., et al. 2012. CNS Neuroscience & Therapeutics.18, 302.
Lynch, G., et al. 2011. Pharmacol. Biochem. Behav.99, 116.
Nakamura, K., et al. 2001. Eur. J. Pharmacol.420, 33.
Ito, I. et al., 1990. J. Physiol.424, 553.
Isaacson et al., 1991. PNAS.88, 1093.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service