Skip to Content
Merck
All Photos(1)

Documents

5.04761

Sigma-Aldrich

CL316243 Hydrate - CAS 138908-40-4 - Calbiochem

Synonym(s):

CL 316,243 hydrate, Disodium 5-[(2R)-2-[[(2R)-2-(3-Chlorophenyl)-2-hydroxyethyl]amino]propyl]-1,3-benzodioxole-2,2-dicarboxylate hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H18ClNNa2O7 · xH2O
CAS Number:
Molecular Weight:
465.79 (anhydrous basis)
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.77

Quality Level

SMILES string

O.[Na+].[Na+].C[C@H](Cc1ccc2OC(Oc2c1)(C([O-])=O)C([O-])=O)NC[C@H](O)c3cccc(Cl)c3

InChI

1S/C20H20ClNO7.2Na.H2O/c1-11(22-10-15(23)13-3-2-4-14(21)9-13)7-12-5-6-16-17(8-12)29-20(28-16,18(24)25)19(26)27;;;/h2-6,8-9,11,15,22-23H,7,10H2,1H3,(H,24,25)(H,26,27);;;1H2/q;2*+1;/p-2/t11-,15+;;;/m1.../s1

InChI key

BFEWLXURNRQQLY-BELDDISZSA-L

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

CL 316,243 is a β3 adrenoceptor agonist; anti-obesity agent.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service