Skip to Content
Merck
All Photos(1)

Key Documents

492899

Sigma-Aldrich

Nuclease, Staphylococcus aureus

Cleaves RNA and DNA to produce oligo- and mononucleotides with terminal 3′-phosphate groups.

Synonym(s):

Nuclease, Staphylococcus aureus

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.54
Pricing and availability is not currently available.

biological source

Staphylococcus aureus

Quality Level

form

lyophilized solid

specific activity

≥15,000 units/mg protein

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

off-white

solubility

aqueous buffer: 1 mg/mL

shipped in

ambient

storage temp.

2-8°C

General description

Native Stapylococcus aureus nuclease. Cleaves RNA and DNA to produce oligo- and mononucleotides with terminal 3′-phosphate groups. Also used to convert standard rabbit reticulocyte cell-free extracts (lysate) into mRNA-dependent protein synthesis systems. Note: 1 KU = 1000 units.

Warning

Toxicity: Standard Handling (A)

Unit Definition

One unit is defined as the amount of enzyme that will release a sufficient amount of acid-soluble oligonucleotides to produce an absorbance increase of 1 at 260 nm in 30 min at 37°C.

Other Notes

Krupp, G., and Gross, H.J. 1979. Nucleic Acids Res.6, 3481.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service