Skip to Content
Merck

Skip To

492002

NOX1 Inhibitor, ML171

The NOX1 Inhibitor, ML171, also referenced under CAS 6631-94-3, controls the biological activity of NOX1.

Synonym(s):

NOX1 Inhibitor, ML171, NADPH Oxidase-1 Inhibitor, ML171, 2-APT, 2-acetylphenothiazine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C14H11NOS
CAS Number:
Molecular Weight:
241.31
UNSPSC Code:
41121801
NACRES:
NA.77
MDL number:
Assay:
≥95% (HPLC)
Form:
powder
Storage condition:
OK to freeze, protect from light
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥95% (HPLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, protect from light

color

dark yellow-orange

solubility

DMSO: 50 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

S1c2c(cc(cc2)C(=O)C)Nc3c1cccc3

InChI

1S/C14H11NOS/c1-9(16)10-6-7-14-12(8-10)15-11-4-2-3-5-13(11)17-14/h2-8,15H,1H3

InChI key

JWGBOHJGWOPYCL-UHFFFAOYSA-N

General description

A highly selective, cell-permeable, and reversible 2-acetylphenothiazine that is shown to inhibit NADPH Oxidase-1 (Nox1) (IC50 = 0.129 µM, and 0.25 µM) in human HT29 and HEK293 cell-based assays, respectively. Unlike most other currently used Nox inhibitors, it only has marginal activity on other cellular ROS-producing enzymes and receptors including the other Nox isoforms (IC50 = 5 µM, 3 µM and 5 µM for Nox2, Nox3, and Nox4, respectively) and xanthine oxidase, XO, (IC50 = 5.5 µM) in HEK293 cell-based assays. It is also shown to block Nox1-dependent ROS generation in HEK293 cultures dose-dependently at concentrations up to ~ 100 nM; however, its inhibitory effect is reversed through over-expression of Nox1, thereby suggesting that this compound is highly selective for Nox1, with little effect against Nox2-dependent ROS generation (IC50 >10 µM). Qualitatively, it displays an inhibitory effect against SrcYF-induced ECM-degrading invadopodia formation in DLD1 Human Colon Cancer Cells.

Packaging

Packaged under inert gas

Preparation Note

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Gianni, D., et al. 2010. ACS Chem. Biol.5, 981.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Regulatory Review (Z)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
219332540213217531
assay

≥95% (HPLC)

assay

≥95% (HPLC)

assay

≥95% (HPLC)

assay

≥95% (HPLC)

form

powder

form

powder

form

solid

form

solid

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

2-8°C

storage condition

OK to freeze, protect from light

storage condition

OK to freeze, protect from light

storage condition

OK to freeze, desiccated (hygroscopic), protect from light

storage condition

OK to freeze, protect from light


Storage Class

11 - Combustible Solids

wgk

WGK 3



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content

"Aging: getting older, exhibiting the signs of age, the decline in the physical (and mental) well-being over time, leading to death. Since the beginning of time, man has been obsessed with trying to slow down, stop, or even reverse the signs of aging. Many have gone as far as experimenting with nutritional regimens, eccentric exercises, fantastic rituals, and naturally occurring or synthetic wonder-elements to evade the signs of normal aging. Biologically speaking, what is aging? And what does the latest research tell us about the possibility of discovering the elusive “fountain of youth”? Many advances in our understanding of aging have come from systematic scientific research, and perhaps it holds the key to immortality. Scientifically, aging can be defined as a systems-wide decline in organismal function that occurs over time. This decline occurs as a result of numerous events in the organism, and these events can be classified into nine “hallmarks” of aging, as proposed by López-Otin et al. (2013). Several of the pathologies associated with aging are a direct result of these events going to extremes and may also involve aberrant activation of proliferation signals or hyperactivity. The hallmarks of aging have been defined based on their fulfillment of specific aging related criteria, such as manifestation during normal aging, acceleration of aging if experimentally induced or aggravated, and retardation of aging if prevented or blocked, resulting in increased lifespan. The nine hallmarks of aging are genomic instability, telomere attrition, epigenetic alterations, loss of proteostasis, deregulated nutrient sensing, mitochondrial dysfunction, cellular senescence, stem cell exhaustion, and altered intercellular communication. The biological processes underlying aging are complex. By understanding the hallmarks in greater detail, we can get closer to developing intervention strategies that can make the aging process less of a decline, and more of a recline."






Global Trade Item Number

SKUGTIN
492002-10MG04055977273465

Questions

Reviews

No rating value

Active Filters