Skip to Content
Merck
All Photos(1)

Key Documents

48712-M

Sigma-Aldrich

o-Nitrophenyl-β-D-galactopyranoside

≥98% (HPLC), solid, β-galactosidase substrate, Calbiochem®

Synonym(s):

o-Nitrophenyl-β-D-galactopyranoside, o-NPG

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H15NO8
CAS Number:
Molecular Weight:
301.25
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Product Name

o-Nitrophenyl-β-D-galactopyranoside, A β-galactosidase substrate for colorimetric and EIA applications; counterpart of widely used pNPP/alkaline phosphatase substrate.

Quality Level

Assay

≥98%

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

impurities

≤0.05% free o-nitrophenol

color

white to off-white

solubility

water: 10 mg/mL

storage temp.

2-8°C

SMILES string

[N+](=O)([O-])c1c(cccc1)OC2OC(C(C(C2O)O)O)CO

InChI

1S/C12H15NO8/c14-5-8-9(15)10(16)11(17)12(21-8)20-7-4-2-1-3-6(7)13(18)19/h1-4,8-12,14-17H,5H2

InChI key

KUWPCJHYPSUOFW-UHFFFAOYSA-N

General description

A β-galactosidase substrate for colorimetric and EIA applications; counterpart of widely used pNPP/alkaline phosphatase substrate. o-Nitrophenol is produced as the end product and is monitored at 405 nm.

Warning

Toxicity: Standard Handling (A)

Other Notes

Maruhn, D. 1976. Clin., Chim. Acta 73, 453.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service