Skip to Content
Merck
All Photos(1)

Key Documents

48-684MAG

Millipore

MILLIPLEX® Ras-Raf Oncoprotein Magnetic Bead Panel 6-Plex - Cell Signaling Multiplex Assay

Synonym(s):

Cell Signaling Assay

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.47
Pricing and availability is not currently available.

species reactivity

human

manufacturer/tradename

Milliplex®

technique(s)

multiplexing: suitable

detection method

fluorometric (Luminex® xMAP®)

storage temp.

2-8°C

General description

Ras proteins are GTPases encoded by three genes: HRAS, KRAS, and NRAS. The protein products of these genes exhibit a high degree of sequence identity and are regulated by a conserved mechanism: exchange of GDP for GTP switches the Ras protein into an active conformation. In its GTP-bound state, Ras has high affinity for its downstream effectors, including the Raf family of protein kinases (ARAF, BRAF, and CRAF/RAF1). Raf proteins, in turn, signal through MEK to activate the MAPK pathway. In healthy cells, Ras activation is a transient response to extracellular growth stimuli and is restricted by the rapid conversion of Ras-GTP back to Ras-GDP. However, the Ras-Raf-MEK pathway is often aberrantly activated in cancer. Mutations in the RAS genes are amongst the most frequently encountered oncogenic events, occurring in ~25% of human cancers, with mutations altering the 12th amino acid (glycine) being particularly prevalent. These driver mutations maintain Ras in a constitutively active, GTP-bound state. Additionally, certain cancers feature KRAS gene amplification, although the effects of gene amplification on protein expression are not yet well-characterized.The MILLIPLEX® MAP 6-plex Ras-Raf Oncoprotein Magnetic Bead Kit (Cat. No. 48-684MAG) is a premixed kit to be used for the simultaneous quantification of the following 6 protein analytes:pBRAF (Ser446)pCRAF (Ser338)pMEK1 (Ser217/Ser221)Ras (total)Mutant Ras protein, RasG12D*Mutant Ras protein, RasG12V**Detects H-Ras, N-Ras, and K-Ras protein

Specificity

Cross-reactivity between the antibodies and any of the other analytes in this panel is non-detectable or negligible.

Application

Intracellular Bead-Based Multiplex Assays using the Luminex technology enable the simultaneous relative quantitation of multiple phosphorylation and total pathway proteins in tissue and cell lysate samples. Compare Multiplexing results to those of Western blotting.An overnight (4°C) incubation is recommended for best results.This kit must be run using Assay Buffer 2 (provided).This assay requires 25 μL diluted cell lysate per well.The suggested working range of protein concentration for the assay is 1 to 25 μg of total protein/well (25 μL/well at 40 to 1,000 μg/mL).Analytes available:BRAF (Ser446);CRAF (Ser338);MEK1 (Ser217/Ser221);Ras (total);RasG12D;RasG12V

Features and Benefits

Configuration: Premixed

Packaging

96-well plate

Storage and Stability

Recommended storage for kit components is 2 - 8°C.

Legal Information

Luminex is a registered trademark of Luminex Corp
MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
xMAP is a registered trademark of Luminex Corp

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Learn how multiplexing with MILLIPLEX® multiplex assays can help simplify cancer research by measuring multiple circulating biomarkers at once. Find multiplex panels to analyze JAK-STAT, mTOR, MAPK, SAPK, TGFβ signaling pathways, and more.

Uncover how cells communicate with MILLIPLEX® cell signaling multiplex assays. Multiplexing with cell signaling phosphoprotein assays based on Luminex® xMAP® technology helps researchers measure phosphoproteins and total proteins within the same or different pathways from a single sample.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service