Skip to Content
Merck
All Photos(1)

Key Documents

4.80508

Supelco

Methanol:Water 30:70 (v:v)

LiChrosolv®

Synonym(s):

30:70 MeOH:H2O solvent

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116105
NACRES:
NA.03
grade:
HPLC grade
technique(s):
HPLC: suitable
application(s):
general analytical
Pricing and availability is not currently available.

grade

HPLC grade

Quality Level

description

isocratic

product line

LiChrosolv®

form

liquid

technique(s)

HPLC: suitable

transition temp

flash point 35 °C

solubility

water: soluble

density

0.95 g/cm3 at 20 °C

application(s)

general analytical

format

mixture

storage temp.

2-30°C

General description

Methanol:Water30:70 (v:v) is a mixture of methanol and water. The key application ofmethnol:water mixture in analytical chemistry as a mobile phase in RPLC(reversed-phase liquid chromatography).[1]

Application

Methanol:Water 30:70 (v:v) can be used as a mobile phase in the HPLC method:
  • For the quantitative determination of 3-hydroxybenzo[a] pyrene and 3-hydroxybenz[a]anthracene in the urine of PAH (polycyclic aromatic hydrocarbon) exposed workers.[2]
  • For the extraction and quantification of hydroxymethylfurfural (HMF) and furfural in bread and bakery products.[3]

Legal Information

LICHROSOLV is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Flam. Liq. 3 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

95.0 °F

Flash Point(C)

35 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Acidity in methanol-water
Rived F, et al.
Analytica Chimica Acta, 439(2), 315-333 (2001)
High-performance liquid chromatographic method with fluorescence detection for the determination of 3-hydroxybenzo [a] pyrene and 3-hydroxybenz [a] anthracene in the urine of polycyclic aromatic hydrocarbon-exposed workers
Gundel J, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 738(1), 47-55 (2000)
Assessment of hydroxymethylfurfural and furfural in commercial bakery products
Petisca C, et al.
Journal of food composition and analysis, 33(1), 20-25 (2014)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service