Skip to Content
Merck
All Photos(1)

Key Documents

480415

Sigma-Aldrich

NF023

A suramin analog that acts as a selective and direct G-protein antagonist for α-subunits of the Go/Gi group (EC₅₀ ~ 300 nM).

Synonym(s):

NF023, 8,8ʹ-[Carbonylbis(imino-3,1-phenylene)]bis-(1,3,5-naphthalenetrisulfonic Acid), 6Na, P2X Antagonist I, Purinergic Receptor P2X Antagonist I

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C35H20N4O21S6 · 6Na
Molecular Weight:
1162.88
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

white to off-white

solubility

water: soluble

shipped in

ambient

storage temp.

2-8°C

General description

A suramin analog that acts as a selective G-protein antagonist for α-subunits of the Go/Gi group (EC50 ~300 nM). Suppresses GTPγS binding to recombinant G-protein α-subunits. Acts by inhibiting the formation of the agonist-specific ternary complex (agonist/receptor/G-protein). NF023 does not interfere with the interaction between α-subunits βγ-dimer but competes with the effector binding site. Also a P2X receptor antagonist (IC50=0.24 µM for P2X1; 8.5 µM for P2X3 receptor in rat muscle).
A suramin analog that acts as a selective and direct G-protein antagonist for α-subunits of the Go/Gi group (EC50 ~ 300 nM). Suppresses GTPγS binding to recombinant G-protein α-subunits. Acts by inhibiting the formation of the agonist-specific ternary complex (agonist/receptor/G-protein). NF023 does not interfere with the interaction between α-subunits and βγ-dimer but competes with the effector binding site. Also a P2X receptor antagonist (IC50 = 240 nM for P2X1; 8.5 µM for P2X3 receptor in rat muscle). Causes a concentration dependent inhibition of excitatory junction potentials (IC50 = 4.8 pM).

Biochem/physiol Actions

Cell permeable: no
Primary Target
α-subunits of the Go/Gi group of G-protein
Product does not compete with ATP.
Reversible: no
Target IC50: 240 nM for P2X1; 8.5 µM for P2X3 receptor in rat muscle; 4.8 pM in the inhibition of excitatory junction potentials; EC50 ~ 300 nM as G-protein antagonist for α-subunits of the Go/Gi group

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Avoid freeze/thaw cycles of solutions.

Other Notes

Snedden, P., et al. 2000. Br. J. Pharmacol.129, 1089.
Soto, F., et al. 1999. Neuropharmacology38, 141.
Beindle, W., et al. 1996. Mol. Pharmacol.50, 415.
Freissmuth, M., et al. 1996. Mol. Pharmacol.49, 602.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service