Skip to Content
Merck
All Photos(1)

Key Documents

475957

Sigma-Aldrich

c-Myc Inhibitor II

The c-Myc Inhibitor II, also referenced under CAS 413611-93-5, controls the biological activity of c-Myc.

Synonym(s):

c-Myc Inhibitor II, 7-Nitro-N-(2-phenylphenyl)-2,1,3-benzoxadiazol-4-amine, Biphenyl-2-yl-(7-nitrobenzo[1,2,5]oxadiazol-4-ylamine, 10074-G5

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H12N4O3
CAS Number:
Molecular Weight:
332.31
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

deep orange

solubility

DMSO: 50 mg/mL

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C18H12N4O3/c23-22(24)16-11-10-15(17-18(16)21-25-20-17)19-14-9-5-4-8-13(14)12-6-2-1-3-7-12/h1-11,19H

InChI key

KMJPYSQOCBYMCF-UHFFFAOYSA-N

General description

A cell-permeable benzoxadiazole compound that is shown to preferentially disrupt the interactions of c-Myc-Max, Mad1-Max, and Myf5-HEB, over those of 31 other pairs of HLH-, ZIP-, and HLH-ZIP-containing proteins. Similarly to 10058-F4 (Cat. No. 475956), 10074-G5 is shown to selectively inhibit the c-Myc-dependent growth of rat fibroblast cell line TGR1 and effectively suppress c-Myc-dependent transcription activity. Binding studies using various mutated and truncated c-Myc constructs reveal that 10074-G5 targets c-Myc helix-1 region between aa residues 363 and 381 (KD = 4.4 µM), while 10058-F4 interaction site is located within aa residues 402 through 412 (KD = 13 µM).

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Follis, A.V., et al. 2008. Chem. Biol.15, 1149.
Yin, X., et al. 2003. Oncogene22, 6151.

Sold under license of U.S. Patent 7,026,343.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yue-Zhi Lee et al.
Pharmacological research, 152, 104581-104581 (2019-12-04)
Interruption of the Warburg effect - the observation that un-stimulated macrophages reprogram their core metabolism from oxidative phosphorylation toward aerobic glycolysis to become pro-inflammatory M1 macrophages upon stimulation - is an emerging strategy for the treatment of cancer and anti-inflammatory
Ashutosh Singh et al.
The Journal of biological chemistry, 297(1), 100903-100903 (2021-06-23)
c-Myc is a transcription factor that plays a crucial role in cellular homeostasis, and its deregulation is associated with highly aggressive and chemotherapy-resistant cancers. After binding with partner MAX, the c-Myc-MAX heterodimer regulates the expression of several genes, leading to
Batuhan Mert Kalkan et al.
Microvascular research, 130, 104001-104001 (2020-03-22)
Endothelial dysfunction is prominent in atherosclerosis, hypertension, diabetes, peripheral and cardiovascular diseases, and stroke. Novel therapeutic approaches to these conditions often involve development of tissue-engineered veins with ex vivo expanded endothelial cells. However, high cell number requirements limit these approaches

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service