Skip to Content
Merck
All Photos(1)

Key Documents

475913

Sigma-Aldrich

Mycophenolic Acid

An immunosuppressive agent that inhibits de novo purine nucleotide synthesis via inhibition of inosine monophosphate dehydrogenase and prevents the formation of XMP and GMP.

Synonym(s):

Mycophenolic Acid, 6-(4-Hydroxy-6-methoxy-7-methyl-3-oxo-5-phthalanyl)-4-methyl-4-hexenoic Acid, MPA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H20O6
CAS Number:
Molecular Weight:
320.34
MDL number:
UNSPSC Code:
12352106
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

description

Merck USA index - 14 6327

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white to off-white

solubility

0.1 M NaOH: 10 mg/mL
ethanol: soluble

shipped in

ambient

storage temp.

10-30°C

SMILES string

O1Cc2c(c(c(c(c2C)OC)C\C=C(\CCC(=O)O)/C)O)C1=O

InChI

1S/C17H20O6/c1-9(5-7-13(18)19)4-6-11-15(20)14-12(8-23-17(14)21)10(2)16(11)22-3/h4,20H,5-8H2,1-3H3,(H,18,19)/b9-4+

InChI key

HPNSFSBZBAHARI-RUDMXATFSA-N

General description

An immunosuppressive agent that inhibits de novo purine nucleotide synthesis via inhibition of inosine monophosphate dehydrogenase and prevents the formation of XMP and GMP. Useful for selection of animal cells that express the E. coli gene coding for xanthine-guanine phosphoribosyltransferase. Also suppresses cytokine-induced nitric oxide production in mouse and rat vascular endothelial cells.
An immunosuppressive agent that inhibits de novo purine nucleotide synthesis via inhibition of IMP dehydrogenase and prevents the formation of XMP and GMP. Useful for selection of animal cells that express the E. coli gene coding for xanthine-guanine phosphoribosyltransferase. Also suppresses cytokine-induced nitric oxide production in mouse and rat vascular endothelial cells.

Biochem/physiol Actions

Cell permeable: no
Primary Target
Inosine monophosphate dehydrogenase
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Harmful & Carcinogenic / Teratogenic (E)

Reconstitution

Following reconstitution, sterilize by filtration through a 0.22 µm pore-size filter, aliquot and freeze (-20°C). Stock solutions are stable for up to 1 month at -20°C.

Other Notes

Langman, L.J., et al. 1995. Clin. Chem. 41, 295.
Senda, M., et al. 1995. Transplantation60, 1143.
Warner-Bartnicki, A.L., et al. 1993. Exp. Cell Res.204, 241.
Falkner, F.G., and Moss, B. 1988. J. Virol.62, 1849.
Cohen, M.B., et al. 1981. J. Biol. Chem.256, 8713.
Mulligan, R.C., and Berg, P. 1981. Proc. Natl. Acad. Sci. USA78, 2072.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Muta. 2 - Repr. 1B - STOT RE 1 Oral

Target Organs

Immune system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Angel K Kongsomboonvech et al.
Frontiers in cellular and infection microbiology, 13, 1130965-1130965 (2023-06-08)
Toxoplasma gondii induces a strong CD8 T cell response characterized by the secretion of IFNγ that promotes host survival during infection. The initiation of CD8 T cell IFNγ responses in vitro differs widely between clonal lineage strains of T. gondii
Alisha Schlichter et al.
eLife, 9 (2020-06-05)
SWI/SNF-family chromatin remodeling complexes, such as S. cerevisiae RSC, slide and eject nucleosomes to regulate transcription. Within nucleosomes, stiff DNA sequences confer spontaneous partial unwrapping, prompting whether and how SWI/SNF-family remodelers are specialized to remodel partially-unwrapped nucleosomes. RSC1 and RSC2

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service