Skip to Content
Merck
All Photos(1)

Key Documents

448103

Sigma-Aldrich

Met/Flt-3/VEGFR2 Tyrosine Kinase Inhibitor

The Met/Flt-3/VEGFR2 Tyrosine Kinase Inhibitor controls the biological activity of Met/Flt-3/VEGFR2 Tyrosine Kinase. Primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

Met/Flt-3/VEGFR2 Tyrosine Kinase Inhibitor, N-(4-(1H-Pyrrolo[2,3-b]pyridin-4-yloxy)-3-fluorophenyl)-1-(4-fluorophenyl)-2-oxo-1,2-dihydropyridine-3-carboxamide, Flt-3 Inhibitor VI, VEGFR Tyrosine Kinase Inhibitor XXVI, VEGFR2 Kinase Inhibitor XXIV

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H16F2N4O3
CAS Number:
Molecular Weight:
458.42
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white to beige
white

solubility

ethanol: 1 mg/mL
DMSO: 10 mg/mL

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C25H16F2N4O3/c26-15-3-6-17(7-4-15)31-13-1-2-19(25(31)33)24(32)30-16-5-8-22(20(27)14-16)34-21-10-12-29-23-18(21)9-11-28-23/h1-14H,(H,28,29)(H,30,32)

InChI key

OBSFXHDOLBYWRJ-UHFFFAOYSA-N

General description

A cell-permeable pyrrolopyridine-pyridone compound that acts as a potent ATP-competitive inhibitor of Met, Flt-3 and VEGFR2 (IC50 = 1.9, 4.0 and 27 nM, respectively), while exhibiting much reduced potency against Lck and c-Kit (IC50 = 290 and 610 nM, respectively) and little or no activity agaisnt Cdk2/CycE, IGF-1R, InsR, MK-2, PKA, and PKCα (IC50 >1 µM). Shown to selectively inhibit the Met-dependent, but not Met-independent, proliferation of gastric carcinoma cultures (IC50 = 60 nM vs. >5 µM against GTL-6 and N87 cells, respectively) in vitro and effectively abolish GTL-6-derived tumor growth (110% reduction of control tumor mass 39 days after tumor implant) by daily oral dosing (50 mg/kg) in mice in vivo.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Kim, K.S., et al. 2008. J. Med. Chem.51, 5330.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service