Skip to Content
Merck
All Photos(1)

Key Documents

444285

Sigma-Aldrich

MMP-2/MMP-9 Inhibitor V - CAS 869577-53-7 - Calbiochem

The MMP-2/MMP-9 Inhibitor V, also referenced under CAS 869577-53-7, controls the biological activity of MMP-2/MMP-9. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.

Synonym(s):

MMP-2/MMP-9 Inhibitor V - CAS 869577-53-7 - Calbiochem, (4-(4-(Methanesulfonamido)phenoxy)phenylsulfonyl)methylthiirane, SB-3CT, pMS

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H17NO5S3
CAS Number:
Molecular Weight:
399.50
UNSPSC Code:
12352200

Quality Level

Assay

≥85% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 200 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

A cell-permeable sulfonamido analog of SB-3CT (Cat. No. 444274) that displays enhanced aqueous solubility and improved selectivity (Ki = 16 nM, 180 nM, and 900 nM for MMP-2, MMP-9, and MMP-14, respectively). Inhibits MMP-3, MMP-7, and MMP-1 only at much higher concentrations (Ki = 3.6 µM, 295 µM, and >25 µM, respectively). Binds to the enzyme active site and acts as an irreversible inhibitor. Reported to potently suppress cell migration and invasion of HT1080 cells (80% inhibition at 2 µM and >25% inhibition at 100 nM, respectively). Unlike the general MMP inhibitor GM6001 (Cat. No. 364205), this inhibitor does not block the activation of cellular pro-MMP-2 by MT1-MMP.

Biochem/physiol Actions

Cell permeable: yes
Product does not compete with ATP.
Reversible: no
Target Ki: 16 nM, 180 nM, and 900 nM for MMP-2, MMP-9, and MMP-14, respectively

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Other Notes

Ikejiri, M., et al. 2005. J. Biol. Chem.280, 33992.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service