442610-M
Malate Dehydrogenase, Porcine Heart
Native malate dehydrogenase from porcine heart. Catalyzes the oxidation of L-malate to oxaloacetate in the presence of NAD+. Note: 1 KU = 1000 units.
Synonym(s):
Malate Dehydrogenase, Porcine Heart, MDH
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
biological source
Porcine heart
Quality Level
form
lyophilized
specific activity
≥100 units/mg dry wt.
≥1000 units/mg protein
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
impurities
≤0.0015% GPT
≤0.01% GOT
≤0.01% LDH
pI
6.1 - 6.4
solubility
0.1 M phosphate buffer: 5 mg/mL
storage temp.
−20°C
General description
Native malate dehydrogenase from porcine heart. Catalyzes the oxidation of L-malate to oxaloacetate in the presence of NAD+. Note: 1 KU = 1000 units.
Native malate dehydrogenase from porcine heart. Catalyzes the oxidation of L-malate to oxaloacetate in the presence of NAD+. Has an optimal pH of 7.2-7.5 and a pI of 6.1-6.4. Note: 1 KU = 1000 units.
Note: 1 KU = 1000 units.
Warning
Toxicity: Standard Handling (A)
Unit Definition
One unit is defined as the amount of enzyme that will oxidize 1.0 µmol NADH per min at 25°C, pH 7.5.
Reconstitution
Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.
Legal Information
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service