Skip to Content
Merck
All Photos(1)

Key Documents

440204

Sigma-Aldrich

LY 294002

InSolution, ≥98%, 10 mM, reversible and specific inhibitor of PI 3-kinase

Synonym(s):

InSolution LY 294002, 2-(4-Morpholinyl)-8-phenyl-4H-1-benzopyran-4-one, BRD2 Inhibitor IV, BRD3 Inhibitor III, BRD4 Inhibitor IV

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H17NO3
CAS Number:
Molecular Weight:
307.34
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (HPLC)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

shipped in

wet ice

storage temp.

−20°C

SMILES string

N4(CCOCC4)c1[o]c2c([c](c1)=O)cccc2c3ccccc3

InChI

1S/C19H17NO3/c21-17-13-18(20-9-11-22-12-10-20)23-19-15(7-4-8-16(17)19)14-5-2-1-3-6-14/h1-8,13H,9-12H2

InChI key

CZQHHVNHHHRRDU-UHFFFAOYSA-N

Biochem/physiol Actions

Cell permeable: yes
Primary Target
phosphatidylinositol 3-kinase
Product competes with ATP.
Reversible: yes
Target IC50: 1.4 µM against phosphatidylinositol 3-kinase

Packaging

Packaged under inert gas

Warning

Toxicity: Irritant (B)

Physical form

A 10 mM (1 mg/325 µl) solution of LY 294002 (Cat. No. 440202) in DMSO.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Other Notes

Dittman, A., et al. 2013. ACS Chem. Biol.9, 495.
Baumann, P., and West, S.C. 1998. Proc. Natl. Acad. Sci. USA95, 14066.
Cardone, M.H., et al. 1998. Science282, 1318.
Vlahos, C.J., et al. 1995. J. Immunol.154, 2413.
Yano, H., et al. 1995. Biochem. J.312, 145.
Vlahos, C.J., et al. 1994. J. Biol. Chem.269, 5241.



Selected Citations
Lee, J., et al. 2009. Cell Stem Cell5, 76.


Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

188.6 °F - (refers to pure substance)

Flash Point(C)

87 °C - (refers to pure substance)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaohua Wu et al.
Molecules (Basel, Switzerland), 26(13) (2021-07-03)
Astragalus membranaceus is a famous herb found among medicinal and food plants in East and Southeastern Asia. The Nrf2-ARE assay-guided separation of an extract from Jing liqueur led to the identification of a nontoxic Nrf2 activator, methylnissolin-3-O-β-d-glucopyranoside (MNG, a component
Abdelkader Essafi et al.
Methods in molecular biology (Clifton, N.J.), 462, 201-211 (2009-01-24)
This chapter describes methods for studying downstream events of the PI3K/Akt signaling cascade, focusing on the FoxO transcription factors. These approaches also represent alternative means for gauging the phosphoinositide-3 kinase/Akt activity. We describe protocols for the fractionation of cytoplasmic and
Anna Bellizzi et al.
Molecular therapy. Nucleic acids, 35(3), 102282-102282 (2024-08-23)
Although our understanding of herpes simplex virus type 1 (HSV-1) biology has been considerably enhanced, developing therapeutic strategies to eliminate HSV-1 in latently infected individuals remains a public health concern. Current antiviral drugs used for the treatment of HSV-1 complications
Christina L Sherry et al.
Journal of immunology (Baltimore, Md. : 1950), 178(2), 663-670 (2007-01-05)
Dysregulated inflammation is a complication of type 2 diabetes (T2D). In this study, we show that augmented LPS-induced TNF-alpha production by resident peritoneal macrophages (PerMphi) in type 2 diabetic (db/db) mice is dependent on elevated glucose and requires p38 MAPK.
Yasuhisa Murai et al.
iScience, 24(10), 103173-103173 (2021-10-26)
Schlafen11 (SLFN11) is referred to as interferon (IFN)-inducible. Based on cancer genomic databases, we identified human acute myeloid and lymphoblastic leukemia cells with gain-of-function mutations in the Janus kinase (JAK) family as exhibiting high SLFN11 expression. In these cells, the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service