Skip to Content
Merck
All Photos(1)

Documents

437629

Sigma-Aldrich

Lipopolysaccharide, Salmonella minnesota Re 595

Stimulates the activity of inducible nitric oxide synthase.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

Quality Level

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

solubility

0.5% triethylamine: soluble

shipped in

ambient

storage temp.

2-8°C

General description

Stimulates the activity of inducible nitric oxide synthase. Induces apoptosis in mouse thymus and swine lymphocytes.

Biochem/physiol Actions

Cell permeable: no
Primary Target
Inducible nitric oxide synthase
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Harmful (C)

Reconstitution

Following reconstitution, store in the refrigerator (4°C). Stock solutions are stable for up to 1 year at 4°C.

Other Notes

Hattori, S., et al. 1995. Biochem. Mol. Biol. Int. 35, 177.
Mayeux, P.R., et al. 1995. Biochem. Pharmacol. 49, 115.
Norimatsu, M., et al. 1995. Infect. Immun.63, 1122.
Feinstein, D.L., et al. 1993. J. Neurochem. 60, 1945.
Zhang, Y.H., et al. 1993. Infect. Immun.61, 5044.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service