Skip to Content
Merck
All Photos(2)

Key Documents

428201

Sigma-Aldrich

LDN-211904

LDN-211904 modulates the biological activity of EphB3 receptor tyrosine kinase. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

LDN-211904, N-(2-chlorophenyl)-6-(piperidin-4-yl)imidazo[1,2-a]pyridine-3-carboxamide oxalate, EphB3 inhibitor, LDN-211904

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H21ClN4O5
CAS Number:
Molecular Weight:
444.87
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥99% (HPLC)

form

solid

potency

79 nM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 50 mg/mL, clear, nearly colorless

shipped in

ambient

storage temp.

2-8°C

SMILES string

O=C(NC1=CC=CC=C1Cl)C2=CN=C3C=CC(C4CCNCC4)=CN32.OC(C(O)=O)=O

InChI

1S/C19H19ClN4O.C2H2O4/c20-15-3-1-2-4-16(15)23-19(25)17-11-22-18-6-5-14(12-24(17)18)13-7-9-21-10-8-13;3-1(4)2(5)6/h1-6,11-13,21H,7-10H2,(H,23,25);(H,3,4)(H,5,6)

InChI key

ODBJGLKPAQMCJA-UHFFFAOYSA-N

General description

A pyrazolo[1,5-a]pyridine derivative that acts as an potent and reversible EphB3 receptor tyrosine kinase inhibitor with an IC50 of 79 nM with good mouse liver microsomal stability and aqueous solubility. Shown to suppress EphB3-induced autophosphorylation of EphB3 receptor at 10 µM in cells. When profiled against a panel of 288 kinases at 5 µM, found to also inhibit most of the Eph (Erythropoietin-producing hepatocellular carcinoma) receptor kinases including EphA1, EphA2, EphA3, EphA4, EphA5, EphA8, EphB1, EphB2, EphB3, EphB4 and other tyrosine kinases, but not EphA6 and EphA7, and is non-inhibitory towards non-RTK kinases screened except for p38α, p38β, and Qik.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Erythropoietin-producing hepatocellular carcinoma (Eph)
Reversible: yes

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Qiao, L., et al. 2011, Bioorg. Med. Chem. Lett.19, 6122

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service