Skip to Content
Merck
All Photos(1)

Key Documents

420126

Sigma-Aldrich

JAK3 Inhibitor VI

The JAK3 Inhibitor VI, also referenced under CAS 856436-16-3, controls the biological activity of JAK3. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

JAK3 Inhibitor VI

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H13N3O · xCH4O3S
CAS Number:
Molecular Weight:
287.32 (free base basis)
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (HPLC)

form

solid

potency

27 nM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

yellow

solubility

water: 1 mg/mL
DMSO: 5 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

[S](=O)(=O)(O)C.[nH]1c(ccc1)\C=C2\c3c(ccc(c3)c4cnccc4)NC\2=O

InChI

1S/C18H13N3O.CH4O3S/c22-18-16(10-14-4-2-8-20-14)15-9-12(5-6-17(15)21-18)13-3-1-7-19-11-13;1-5(2,3)4/h1-11,20H,(H,21,22);1H3,(H,2,3,4)/b16-10-;

InChI key

ZUDPNBZMAJGYTE-HYMQDMCPSA-N

General description

A cell-permeable 3′-pyridyl oxindole compound that acts as a potent and reversible inhibitor of JAK3 (IC50 = 27 nM)and displays ~16-fold greater selectivity over JAK2. Binds to the enzyme active site and prevents IL-2-induced cellular phosphorylation of JAK3 and STAT5. Inhibits IL-2-induced cell proliferation (IC50 = 760 and 250 nM for mouse CTLL and human T-cells, respectively) and alleviates oxazolone-induced ear odema in mouse with an efficacy comparable to that of Dexamethasone (Cat. No. 265005).
A cell-permeable, potent, and reversible inhibitor of Janus kinase 3 (JAK3) (IC50 = 27 nM). Reported to bind to the enzyme active site and prevent IL-2-induced phosphorylation of JAK3 and STAT5. Also reported to inhibit IL-2-induced cell proliferation (IC50 = 760 nM for murine CTLL cells and 250 nM for human T-cells). Shown to alleviate oxazolone-induced ear edema in mice with an efficacy comparable to Dexamethasone (Cat. No. 265005) and displays ~16-fold greater selectivity over JAK2.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
JAK-3
Product does not compete with ATP.
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Carcinogenic / Teratogenic (D)

Reconstitution

Following reconstitution aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Chen, J., et al. 2006. Bioorg. Med. Chem. Lett. In press.
Adams, C., et al. 2003. Bioorg. Med. Chem. Lett.13, 3105.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sean M Buchanan et al.
Skeletal muscle, 10(1), 28-28 (2020-10-11)
Satellite cells are the canonical muscle stem cells that regenerate damaged skeletal muscle. Loss of function of these cells has been linked to reduced muscle repair capacity and compromised muscle health in acute muscle injury and congenital neuromuscular diseases. To

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service