Skip to Content
Merck
All Photos(1)

Key Documents

407707

Sigma-Aldrich

PhosphoDetect Anti-Insulin Receptor (pTyr1162/1163) Rabbit pAb

liquid, Calbiochem®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

antibody product type

primary antibodies

clone

polyclonal

form

liquid

contains

≤0.1% sodium azide as preservative

species reactivity

human

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG

shipped in

wet ice

storage temp.

−70°C

target post-translational modification

phosphorylation (pTyr1162/pTyr1163)

Gene Information

human ... INSR(3643)

General description

Note: 1 T = 1 test. Sufficient for 10 Western mini blots.
Rabbit polyclonal antibody adsorbed with non-phosphopeptide corresponding to the immunogen phosphorylation site, followed by immunoaffinity chromatography. Recognizes the insulin receptor (IR) protein phosphorylated at Tyr1162/1163 and insulin-like growth factor 1 receptor (IGF-1R) protein phosphorylated at Tyr1162/1163.
Recognizes the ~95 kDa β subunit of the insulin receptor protein phosphorylated at Tyr1162 and Tyr1163 in insulin receptor transfected cells. Cross-reacts with the insulin-like growth factor-1 receptor phosphorylated at Tyr1135 and Tyr1136.
This PhosphoDetect Anti-Insulin Receptor (pTyr1162/1163) Rabbit pAb is validated for use in ELISA, WB, ICC for the detection of Insulin Receptor (pTyr1162/1163).

Immunogen

Human
a synthetic phosphopeptide corresponding to amino acids surrounding the Tyr1162/1163 phosphorylation sites of human insulin receptor

Application

ELISA (see comments)

Immunoblotting (1:1000)

Immunocytochemistry (see comments)

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Other Notes

Bevan, P. 2001. J. Cell Sci.114, 1429.
Ottensmeyer, F.P., et al. 2000. Biochemistry39, 12103.
Virkamaki, A., et al. 1999. J. Clin. Invest.103, 931.
Schmid, E., at al. 1998. FASEB J.12, 863.
Hari, J. and Roth, R.A. 1987. J. Biol. Chem.262, 15341.
Morgan, D.O., et al. 1986. Proc. Natl. Acad. Sci. USA83, 328.
White, M.F. and Kahn, C.R. 1986. The Enzymes17, 247.
Ebina, Y., et al. 1985. Cell40, 747.
Ganguly, S., et al. 1985. Current Topics in Cellular Regulation27, 83.
Grunberger, G., et al. 1984. Science223, 932.
The immunogen sequence is 100% conserved in the mouse and rat insulin recptor protein, but cross-reactivity has not been tested. This antibody has also been reported to work for ELISA and immunocytochemistry. Variables associated with assay conditions will dictate the proper working dilution.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tatsuya Iso et al.
Arteriosclerosis, thrombosis, and vascular biology, 33(11), 2549-2557 (2013-08-24)
Fatty acids (FAs) are the major substrate for energy production in the heart. Here, we hypothesize that capillary endothelial fatty acid binding protein 4 (FABP4) and FABP5 play an important role in providing sufficient FAs to the myocardium. Both FABP4/5
Ana S Martins et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 12(11 Pt 1), 3532-3540 (2006-06-03)
Ewing tumor cell survival and proliferation depends on several autocrine loops. Targeting these loops is a promising therapeutic approach. We recently showed the cytostatic role of imatinib, an inhibitor of the SCF-KIT loop, on Ewing tumor cells, and in this
Ekin Guney et al.
Science signaling, 14(713), eabf2059-eabf2059 (2021-12-15)
Chronic metabolic inflammation is a key feature of obesity, insulin resistance, and diabetes. Here, we showed that altered regulation of the Ca2+ channel inositol trisphosphate receptor (IP3R) was an adipocyte-intrinsic event involved in the emergence and propagation of inflammatory signaling
Natalie J Haywood et al.
Diabetes, 66(2), 287-299 (2017-01-22)
Low circulating levels of insulin-like growth factor binding protein 1 (IGFBP-1) are associated with insulin resistance and predict the development of type 2 diabetes. IGFBP-1 can affect cellular functions independently of IGF binding through an Arg-Gly-Asp (RGD) integrin-binding motif. Whether
Ana Paula Arruda et al.
eLife, 6 (2017-12-16)
Defective Ca2+ handling is a key mechanism underlying hepatic endoplasmic reticulum (ER) dysfunction in obesity. ER Ca2+ level is in part monitored by the store-operated Ca2+ entry (SOCE) system, an adaptive mechanism that senses ER luminal Ca2+ concentrations through the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service