Skip to Content
Merck
All Photos(1)

Documents

407601

Sigma-Aldrich

Interleukin-1 Receptor-Associated-Kinase-1/4 Inhibitor

The Interleukin-1 Receptor-Associated-Kinase-1/4 Inhibitor controls the activity of Interleukin-1 Receptor-Associated-Kinase-1/4. Primarily used for Phosphorylation & Dephosphorylation apps.

Synonym(s):

Interleukin-1 Receptor-Associated-Kinase-1/4 Inhibitor, IRAK-1/4 Inhibitor, N-(2-Morpholinylethyl)-2-(3-nitrobenzoylamido)-benzimidazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H21N5O4
CAS Number:
Molecular Weight:
395.41
UNSPSC Code:
12352202
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

crystalline solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 10 mg/mL

shipped in

ambient

storage temp.

−20°C

InChI

1S/C20H21N5O4/c26-19(15-4-3-5-16(14-15)25(27)28)22-20-21-17-6-1-2-7-18(17)24(20)9-8-23-10-12-29-13-11-23/h1-7,14H,8-13H2,(H,21,22,26)

InChI key

QTCFYQHZJIIHBS-UHFFFAOYSA-N

General description

A cell-permeable benzimidazole compound that acts as a potent and selective inhibitor of interleukin-1 receptor-associated kinases (IC50 = 300 nM and 200 nM for IRAK-1 and -4). Shown to exhibit little activity against a panel of 27 other kinases (IC50 >10 µM), including lck and src.
A cell-permeable benzimidazole compound that acts as a potent and selective inhibitor of interleukin-1 receptor-associated kinases (IC50 = 300 nM and 200 nM for IRAK-1 and -4). Shown to exhibit little activity against a panel of 27 other kinases (IC50 >10 µM), including lck and src. Also available as a 25 mM solution in DMSO (Cat. No. 407602).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Interleukin-1 receptor-associated kinases
Product does not compete with ATP.
Reversible: no
Target IC50: 300 nM and 200 nM for IRAK-1 and -4

Packaging

Packaged under inert gas

Warning

Toxicity: Harmful (C)

Other Notes

Bhattacharyya, S., et al. 2007. Am. J. Physiol. Gastrointest. Liver Physiol.293, G429.
Powers, J.P., et al. 2006. Bioorg. Med. Chem. Lett.16, 2842.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Steven P Treon et al.
The New England journal of medicine, 367(9), 826-833 (2012-08-31)
Waldenström's macroglobulinemia is an incurable, IgM-secreting lymphoplasmacytic lymphoma (LPL). The underlying mutation in this disorder has not been delineated. We performed whole-genome sequencing of bone marrow LPL cells in 30 patients with Waldenström's macroglobulinemia, with paired normal-tissue and tumor-tissue sequencing

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service