Skip to Content
Merck
All Photos(1)

Key Documents

401504

Sigma-Aldrich

Rabbit Anti-Goat IgG, Fc Fragment Specific Peroxidase Conjugate

liquid, Calbiochem®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

liquid

contains

0.02% thimerosal as preservative

manufacturer/tradename

Calbiochem®

storage condition

do not freeze

isotype

IgG

shipped in

wet ice

storage temp.

2-8°C

General description

Adsorbed against bovine, horse, and human serum proteins.
Immunoaffinity purified rabbit polyclonal antibody adsorbed against bovine, horse, and human serum proteins and conjugated to horseradish peroxidase (HRP). Recognizes goat IgG Fc fragment.
This Rabbit Anti-Goat IgG, Fc Fragment Specific Peroxidase Conjugate is validated for use in Enzyme Immunoassay, Immunoelectrophoresis for the detection of Goat IgG, Fc Fragment Specific.

Immunogen

Goat

Application

Enzyme Immunoassay (1:10,000)

Immunoelectrophoresis (see comments)

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

in 10 mM PBS, 0.5 mM EDTA, 10% mannose, 2% sucrose, 1% BSA, pH 7.6.

Other Notes

Monospecific for goat IgG Fc fragment as determined by immunoelectrophoresis against normal goat serum. o-Phenylenediamine, 1 mg/ml in citrate buffer, pH 4.5, 0.1% urea peroxide at 25°C, was used as the substrate for testing immunoenzyme reactivity. Reactivity with normal bovine, horse, and human serum is ≤2% by direct solid phase immunoassay. Variables associated with assay conditions will dictate the proper working dilution.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

To comply with ban of sale of mercury-added products required by The Interstate Mercury Education and Reduction Clearinghouse (IMERC), this product is prohibited to be sold in the following US states: Rhode Island and Connecticut.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Antonio Ferreira Mendes-Sousa et al.
PloS one, 8(11), e79787-e79787 (2013-11-21)
Lutzomyia longipalpis is the vector of Leishmania infantum in the New World, and its saliva inhibits classical and alternative human complement system pathways. This inhibition is important in protecting the insect´s midgut from damage by the complement. L. longipalpis is
U Schneider et al.
Journal of virology, 74(21), 9928-9936 (2000-10-12)
We sought proof of principle that one of the safest human vaccines, measles virus Edmonston B (MV-Edm), can be genetically modified to allow entry via cell surface molecules other than its receptor CD46. Hybrid proteins consisting of the epidermal growth
Sini Heinonen et al.
Diabetologia, 60(1), 169-181 (2016-10-14)
Low mitochondrial activity in adipose tissue is suggested to be an underlying factor in obesity and its metabolic complications. We aimed to find out whether mitochondrial measures are downregulated in obesity also in isolated adipocytes. We studied young adult monozygotic
Veruska Cavalcanti Barros et al.
PloS one, 4(6), e6047-e6047 (2009-06-27)
Saliva of haematophagous arthropods contain biomolecules involved directly or indirectly with the haematophagy process, and among them are encountered some complement system inhibitors. The most obvious function for these inhibitors would be the protection of the midgut against injury by
Georgina Espígol-Frigolé et al.
Science signaling, 9(419), ra28-ra28 (2016-03-17)
Interleukin-23 (IL-23), a heterodimeric cytokine composed of the unique p19 peptide (IL-23p19) and a peptide called IL-12p40, which is shared with IL-12, is implicated in Crohn's disease, rheumatoid arthritis, psoriasis, and other immune-mediated inflammatory diseases. Endothelial cells produce the IL-23p19

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service