401483
IKK-2 Inhibitor VI - CAS 354811-10-2 - Calbiochem
Synonym(s):
(5-Phenyl-2-ureido)thiophene-3-carboxamide
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
General description
An ureido-thiophenecarboxamide compound that acts as a potent, cell-permeable, reversible, and ATP-competitive inhibitor of IKK-2 (IC50 = 13 nM).
Biochem/physiol Actions
IKK-2
Reconstitution
Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.
Storage Class Code
10-13 - German Storage Class 10 to 13
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Andrew Baxter et al.
Bioorganic & medicinal chemistry letters, 14(11), 2817-2822 (2004-05-06)
A hit-to-lead optimisation programme was carried out on the thiophenecarboxamide high throughput screening hits 1 and 2 resulting in the discovery of the potent and orally bioavailable IKK-2 inhibitor 22.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service