401479
IKK-2 Inhibitor, SC-514
The IKK-2 Inhibitor, SC-514, also referenced under CAS 354812-17-2, controls the biological activity of IKK-2. This small molecule/inhibitor is primarily used for Inflammation/Immunology applications.
Synonym(s):
IKK-2 Inhibitor, SC-514, SC-514
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C9H8N2OS2
CAS Number:
Molecular Weight:
224.30
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Recommended Products
Quality Level
Assay
≥95% (HPLC)
form
solid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
protect from light
color
pale yellow
solubility
DMSO: 20 mg/mL
shipped in
ambient
storage temp.
2-8°C
SMILES string
[s]1c(cc(c1C(=O)N)N)c2c[s]cc2
InChI
1S/C9H8N2OS2/c10-6-3-7(5-1-2-13-4-5)14-8(6)9(11)12/h1-4H,10H2,(H2,11,12)
InChI key
BMUACLADCKCNKZ-UHFFFAOYSA-N
General description
A cell-permeable (thienothienyl)amino-acetamide compound that displays anti-inflammatory properties. Acts as a potent, reversible, ATP-competitive, and highly selective inhibitor of IKK-2 (IC50 = ~ 3-12 µM for IKK-2 homodimer, IKK-1/IKK-2 heterodimer, and IKK-2). Its specificity has been confirmed using a panel of 31 other kinases, including IKK isoforms IKK-1, IKK-i, and TBK-1 (IC50 >200 µM). Shown to specifically block NF-κB-dependent gene expression, but not MAP kinase pathways, in stimulated synovial fibroblasts RASF. A 25 mM (1 mg/178 µl) solution of IKK-2 Inhibitor, SC-514 (Cat. No. 401485) in DMSO is also available.
A cell-permeable, potent, reversible, ATP-competitive and highly selective inhibitor of IKK-2 (IC50 ~3-12 µM for IKK-2 homodimer, IKK-1/IKK-2 heterodimer, and IKK-2) that also displays anti-inflammatory properties. Specificity has been reported using a panel of 31 additional kinases, including IKK isoforms IKK-1, IKK-i, and TBK-1 (IC50 >200 µM). Shown to specifically block NF-κB-dependent gene expression, but not MAP kinase pathways, in stimulated RASF synovial fibroblast cells. Does not inhibit the phosphorylation and activation of the IKK complex.
Biochem/physiol Actions
Cell permeable: yes
Primary Target
IKK-2
IKK-2
Product competes with ATP.
Reversible: yes
Target IC50: 3-12 µM for IKK-2 homodimer, IKK-1/IKK-2 heterodimer, and IKK-2
Packaging
Packaged under inert gas
Warning
Toxicity: Carcinogenic / Teratogenic (D)
Reconstitution
Following reconstitution aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.
Other Notes
Baxter, A., et al. 2004 Bioorg. Med. Chem. Lett.14, 2817.
Kishore, N., et al. 2003. J. Biol. Chem.278, 32861.
Kishore, N., et al. 2003. J. Biol. Chem.278, 32861.
Legal Information
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Zahara L Chaudhry et al.
Brain sciences, 10(11) (2020-11-05)
The current coronavirus pandemic caused by the severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2) has resulted in a serious global health crisis. It is a major concern for individuals living with chronic disorders such as Parkinson's disease (PD). Increasing evidence
Maike Mose et al.
Experimental dermatology, 22(7), 476-481 (2013-06-27)
The antimicrobial peptide S100A8 is known to be upregulated in lesional psoriatic skin compared with non-lesional psoriatic skin and is believed to play a role in the pathogenesis of psoriasis. However, little is known about the signalling pathways involved in
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service