Skip to Content
Merck
All Photos(1)

Documents

400086

Sigma-Aldrich

HIF Inhibitor VI

The HIF-1 Inhibitor, NSC-134754 controls the biological activity of HIF-1. This small molecule/inhibitor is primarily used for Cell Structure applications.

Synonym(s):

HIF Inhibitor VI, NSC-134754

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H34N2O2
Molecular Weight:
418.57
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥90% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

brown

solubility

DMSO: 50 mg/mL

shipped in

ambient

storage temp.

2-8°C

General description

A cell-permeable emetan derivative that selectively reduces HIF-1 activity by 73% in a luciferase reporter assay, and blocks HIF-1α protein induction in response to DFX, hypoxia, and IGF-1 in U2OS-HRE-luc cells at 10 µM, without significantly inhibiting cell growth. Furthermore, it is shown to completely block HIF-1α induced by IGF-1 in HCT116 human colon carcinoma cells.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Other Notes

Chau, N.M., et al. 2005. Cancer Res.65, 4918.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service