Skip to Content
Merck
All Photos(1)

Key Documents

393206

Sigma-Aldrich

Anti-HNE Fluorophore Rabbit pAb

liquid, Calbiochem®

Synonym(s):

Anti-N α-Acetyllysine-4-hydroxy-2-nonenal Fluorophore, Anti-Nα-Acetyllysine-4-hydroxy-2-nonenal Fluorophore

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

antibody form

diluted serum

antibody product type

primary antibodies

clone

polyclonal

form

liquid

species reactivity (predicted by homology)

all

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG

shipped in

wet ice

storage temp.

−70°C

target post-translational modification

unmodified

General description

Anti-HNE Fluorophore, rabbit polyclonal, recognizes the HNE-derived chromphohore of the 2:1 lysine-HNE-fluorophore. It is validated for use in ELISA, Western blotting, and immunohistochemistry.
Rabbit polyclonal antibody supplied as diluted serum. Recognizes the HNE-derived chromophore of the 2:1 lysine-HNE fluorophore.
Recognizes the HNE-derived chromphohore of the 2:1 lysine-HNE-fluorophore.

Immunogen

a 2:1 Nα-acetyllysine-HNE fluorophore, conjugated to KLH

Application

ELISA (1:500-1:1000)

Immunoblotting (1:1000-1:5000)

Immunohistochemistry (1:100-1:500)

Warning

Toxicity: Standard Handling (A)

Physical form

Serum diluted 1:1 with glycerol.

Reconstitution

Following initial thaw, aliquot and freeze (-70°C).

Other Notes

Due to the small epitope size, overnight incubation with the antibody greatly increases sensitivity by immunoblotting. Determined by competitive ELISA and immunoblotting to recognize the chromophore structure of fluorophore derived from the reaction of 4-HNE with primary amines. Does not recognize 1:1 cysteine-, 1:1 histidine-, or 1:1 lysine-HNE Michael Adducts. Variables associated with assay conditions will dictate the proper working dilution.
Tsai, L., et al. 1998. Proc. Natl. Acad. Sci. USA95, 7975.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ernst-Bernhard Kayser et al.
PloS one, 11(1), e0148219-e0148219 (2016-01-30)
Lack of NDUFS4, a subunit of mitochondrial complex I (NADH:ubiquinone oxidoreductase), causes Leigh syndrome (LS), a progressive encephalomyopathy. Knocking out Ndufs4, either systemically or in brain only, elicits LS in mice. In patients as well as in KO mice distinct
Mara Laslo et al.
Age (Dordrecht, Netherlands), 35(4), 1117-1132 (2012-05-29)
Superoxide dismutase 1 (SOD1), a critical enzyme against oxidative stress, is implicated in aging and degenerative diseases. We previously showed that a nutraceutical containing freeze-dried açai pulp promotes survival of flies fed a high-fat diet or sod1 knockdown flies fed
Erik P Rader et al.
Rejuvenation research, 20(2), 93-102 (2016-07-06)
Exercise is the most accessible, efficacious, and multifactorial intervention to improve health and treat chronic disease. High-intensity resistance exercise, in particular, also maximizes skeletal muscle size and strength-outcomes crucial at advanced age. However, such training is capable of inducing muscle
Olga Boyd et al.
Free radical biology & medicine, 50(11), 1669-1678 (2011-03-17)
Fruits containing high antioxidant capacities and other bioactivities are ideal for promoting longevity and health span. However, few fruits are known to improve the survival and health span in animals, let alone the underlying mechanisms. Here we investigate the effects
Marshall A Naimo et al.
Journal of applied physiology (Bethesda, Md. : 1985), 126(4), 1074-1087 (2019-01-25)
The purpose of this study was to characterize the growth and remodeling molecular signaling response in aged skeletal muscle following 1 mo of "resistance-type exercise" training. Male Fischer 344 × Brown Norway hybrid rats aged 3 (young) and 30 mo

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service