Skip to Content
Merck
All Photos(1)

Documents

391947

Sigma-Aldrich

Hydroxylapatite, Fast Flow

Synonym(s):

Hydroxylapatite, Fast Flow, HA

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41116133
NACRES:
NA.77

form

solid

Quality Level

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

phosphate buffer: soluble

shipped in

ambient

storage temp.

10-30°C

InChI

1S/5Ca.3H3O4P.H2O/c;;;;;3*1-5(2,3)4;/h;;;;;3*(H3,1,2,3,4);1H2/q5*+2;;;;/p-10

InChI key

XYJRXVWERLGGKC-UHFFFAOYSA-D

General description

A form of calcium phosphate used in preparative procedures for purification of proteins and nucleic acids in which high flow rate and binding capacity are desired (BSA binding capacity: >12 mg/g; DNA binding capacity: >700 µg/g).
A form of calcium phosphate used in preparative procedures for purification of proteins and nucleic acids in which high flow rate and binding capacity are desired. BSA binding capacity: >12 mg/g; DNA binding capacity: >700 µg/g.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, store in the refrigerator (4°C).

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service