Skip to Content
Merck
All Photos(1)

Documents

386445

Sigma-Aldrich

Hybridoma Subisotyping Kit, Mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116133
NACRES:
NA.41

manufacturer/tradename

Calbiochem®

Quality Level

storage condition

OK to freeze

storage temp.

2-8°C

General description

The Hybridoma Subisotyping Kit is intended for the detection and identification of mouse monoclonal antibody subclasses IgG1, IgG2a, IgG2b, IgM and IgA in supernatants. It is also used for confirmation of monoclonality of cloned hybridoma cultures. This double antibody detection system is a highly sensitive enzyme immunoassay method incorporating specific primary antisera. Assay sensitivity is less than 1 µg/ml and can be reliably used with diluted culture supernatants. The following protocol utilizes 50 µl X 7 (350 µl) of each supernatant tested (a 1:2 dilution occurs as each sample is added to diluent in each well). When supernatant medium is precious, the medium from high producer cells can be diluted much more (1:10 or greater), with appropriate modifications in the Application of Supernatants section. When used as directed, the reagents in this kit will be sufficient for 900 determinations or 150 clones
Useful to confirm the purity of cell lines in monoclonal antibody development assays. Sufficient to perform 900 assays (150 clones).

Components

Antibodies to IgA, IgG₁, IgG2a, IgG2b, IgG₃, IgM, anti-mouse peroxidase conjugate, TMB substrate solution, PBS concentrate, surfactant, and a user protocol.

Warning

Toxicity: Multiple Toxicity Values, refer to MSDS (O)

Storage and Stability

Upon arrival store the entire contents of the kit at 4°C.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hua Li et al.
Molecular therapy : the journal of the American Society of Gene Therapy, 17(7), 1215-1224 (2009-04-16)
Hepatic adeno-associated virus (AAV)-serotype 2-mediated gene transfer results in sustained transgene expression in experimental animals but not in human subjects. We hypothesized that loss of transgene expression in humans might be caused by immune memory mechanisms that become reactivated upon

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service