Skip to Content
Merck
All Photos(2)

Key Documents

382115

Sigma-Aldrich

Histone Acetyl Transferase Inhibitor VII, CTK7A

The Histone Acetyl Transferase Inhibitor VII, CTK7A controls the biological activity of Histone Acetyl Transferase. This small molecule/inhibitor is primarily used for Cell Structure applications.

Synonym(s):

Histone Acetyl Transferase Inhibitor VII, CTK7A, HAT Inhibitor VII, p300/CBP Inhibitor V, PCAF Inhibitor III, Sodium-4-(3,5-bis(4-hydroxy-3-methoxystyryl)-1H-pyrazol-1-yl)benzoate, HBC, Na, Sodium-4-(3,5-bis(4-hydroxy-3-methoxystyryl)-1H-pyrazol-1-yl)benzoate, HBC, Na, HAT Inhibitor VII, p300/CBP Inhibitor V, PCAF Inhibitor III

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H23N2NaO6
Molecular Weight:
506.48
UNSPSC Code:
12352200
NACRES:
NA.32
Pricing and availability is not currently available.

Quality Level

Assay

≥99% (HPLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

tan

solubility

DMSO: 100 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

OC1=CC=C(C=C1OC)/C=C/C2=CC(/C=C/C3=CC=C(O)C(OC)=C3)=NN2C4=CC=C(C=C4)C(O[Na])=O

General description

A cell-permeable and water soluble hydrazinobenzoylcurcumin (HBC) compound that acts as a selective, reversible and mixed mode inhibitor of p300/CBP and PCAF histone acetyltransferases (IC50 ≤ 25 and 50 µM, respectively) with minimal activity towards other histone modifying enzymes like, G9a, CARM1, HDAC1, SIRT2 and TIP60 (IC50 >100 µM). Blocks the autoacetylation of p300 and PCAF and histone acetylation, downregulates cyclin E expression and induces senescence-like growth arrest at 100 µM in KB cells. Further, suppresses tumor growth in xenograft oral tumor nude mouse model (100 mg/kg, i.p., b.i.d.) and reduces H3K14 and H3K9 acetylation levels. Reported to potently inhibit bFGF-induced bovine aortic endothelial cells (BAECs) invasion and tube formation with an IC50 of 0.93 µM and binds to Ca2+/calmodulin with high-affinity (Kd = 8.11 µM) and antagonize its functions in HCT15 cells.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Other Notes

Arif, M., et al. 2010. Chem. Biol.17, 903.
Shim, J.S., et al. 2004. Chem. Biol.11, 1455.
Shim, J.S., et al. 2002. Bioorg. Med. Chem. Biol.10, 2439.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vignesh Ramesh et al.
EMBO molecular medicine, 15(12), e17836-e17836 (2023-09-28)
The epithelial-to-mesenchymal transition (EMT) plays a central role in the development of cancer metastasis and resistance to chemotherapy. However, its pharmacological treatment remains challenging. Here, we used an EMT-focused integrative functional genomic approach and identified an inverse association between short-chain
Johanna Diener et al.
Nature communications, 12(1), 5056-5056 (2021-08-22)
Melanoma cells rely on developmental programs during tumor initiation and progression. Here we show that the embryonic stem cell (ESC) factor Sall4 is re-expressed in the Tyr::NrasQ61K; Cdkn2a-/- melanoma model and that its expression is necessary for primary melanoma formation.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service