Skip to Content
Merck
All Photos(1)

Documents

3745

Sigma-Aldrich

Hemoglobin, Bovine Erythrocytes

Hemoglobin, Bovine Erythrocytes, CAS 9008-02-0, is an iron-containing oxygen-transport metalloprotein. Suitable as a substrate for proteases, such as papain, pepsin, and trypsin.

Synonym(s):

Hemoglobin, Bovine Erythrocytes

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.25

Quality Level

Assay

≥95% (SDS-PAGE)

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

solubility

aqueous buffer: 1 mg/mL
water: 1 mg/mL

shipped in

wet ice

storage temp.

−20°C

General description

Native hemoglobin from bovine erythrocytes. A major oxygen-transporting component of red blood cells that is also nitric oxide scavenger. Blocks carbachol-stimulated cGMP production. This preparation contains primarily Ferric-hemoglobin and must be reduced to the ferrous form to bind molecular oxygen. Note: this preparation contains primarily ferric-hemoglobin and must be reduced to the ferrous form to bind molecular oxygen.
Native hemoglobin from bovine erythrocytes. A major oxygen-transporting component of red blood cells. A nitric oxide scavenger that complexes with nitric oxide. Blocks carbachol-stimulated cGMP production. This preparation contains primarily ferric-hemoglobin and must be reduced to the ferrous form to bind molecular oxygen.

Application

Hemoglobin, Bovine Erythrocytes, CAS 9008-02-0, is an iron-containing oxygen-transport metalloprotein. Suitable as a substrate for proteases, such as papain, pepsin, and trypsin.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following recosntitution aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Gow, A.J., et al. 1999. Proc. Natl. Acad. Sci. USA 96, 9027.
Castoldi, A.F., et al. 1993. Brain Res.610, 57.
Bredt, D.S., and Snyder, S.H. 1989. Proc. Natl. Acad. Sci. USA86, 9030.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service