Skip to Content
Merck
All Photos(1)

Key Documents

362331

Sigma-Aldrich

GAG Agonist, Adhesamine - Calbiochem

Synonym(s):

Glycosaminoglycans Agonist

Sign Into View Organizational & Contract Pricing


About This Item

Assay

≥80% (HPLC)

form

solid

color

light yellow

solubility

DMSO: 25 mg/mL

General description

A symmetrical nonpeptidyl cationic diaryldispirotripiperazine compound that binds GAGs (glycosaminoglycans) in a reversible manner (KD = 0.39, 4.67, and 5.85 μM for heparin, heparan sulfate, and keratin sulfate, respectively) and effectively promotes adhesion (by ~100 and 600% at 60 μM, respectively) and growth (by ~80 and 50% in 72 h at 1 μM, respectively) of HepG2 and Jurkat cells in a dose-dependent manner. Binding studies using heparin-derived oligosaccharides indicate that the major mode of interaction involves the binding of a hexasaccharide unit by two adhesamine molecules (KD = 0.12 μM). Primary murine hippocampal neurons cultured in the presence of adhesamine are reported to exhibit earlier differentiation as well as improved survival rate when compared with neurons cultured on poly-L-lysine-coated surface. A GAGs binding antagonist Surfen is also available under Cat. No. 362330.
The GAG Agonist, Adhesamine controls the biological activity of GAG. This small molecule/inhibitor is primarily used for Activators/Inducers applications.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 1 month at -20°C.

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mitsunobu Hoshino et al.
The Biochemical journal, 427(2), 297-304 (2010-02-12)
Attachment to the substrate is essential for both survival and differentiation of various kinds of cells, such as neurons and epithelial cells. We recently found a small synthetic molecule, adhesamine, which boosts adhesion and growth of mammalian cells. In the
Sayumi Yamazoe et al.
Chemistry & biology, 16(7), 773-782 (2009-07-29)
During an image-based phenotype screening of our chemical library, we noted a small molecule that boosts the adhesion and growth of human cells. Chemical and cell biological experiments suggest that the diaryldispirotripiperazine derivative (adhesamine) targets selective cell-surface glycosaminoglycans, especially heparan

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service