Skip to Content
Merck
All Photos(1)

Key Documents

361554

Sigma-Aldrich

TWS119

≥95% (HPLC), solid, GSK-3β inhibitor, Calbiochem®

Synonym(s):

GSK-3β Inhibitor XII, TWS119, Neurogenesis Inducer, TWS119

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H14N4O2
CAS Number:
Molecular Weight:
318.33
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Product Name

GSK-3β Inhibitor XII, TWS119, The GSK-3β Inhibitor XII, TWS119, also referenced under CAS 601514-19-6, controls the biological activity of GSK-3β. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Quality Level

Assay

≥95% (HPLC)

form

solid

potency

30 nM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

beige

solubility

methanol: 2 mg/mL
DMSO: 200 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

[nH]1c2nc(cc2c(nc1)Oc4cc(ccc4)O)c3cc(ccc3)N

InChI

1S/C18H14N4O2/c19-12-4-1-3-11(7-12)16-9-15-17(22-16)20-10-21-18(15)24-14-6-2-5-13(23)8-14/h1-10,23H,19H2,(H,20,21,22)

InChI key

VPVLEBIVXZSOMQ-UHFFFAOYSA-N

General description

A cell-permeable and potent inhibitor of GSK-3β (IC50 = 30 nM). Binds to GSK-3β with high-affinity (Kd = 126 nM), leading to an increase in the level of β-catenin, a downstream substrate of GSK-3β in the Wnt signaling pathway. Shown to selectively induce neuronal differentiation in both pluripotent murine embryonal carcinoma cells (P19; 30-40% at 1 µM) and embryonic stem cells (D3; 50-60% at 400 nM).
A cell-permeable pyrrolopyrimidine compound that acts as a potent inhibitor of GSK-3β (IC50 = 30 nM). Binds to GSK-3β with high-affinity (Kd = 126 nM) and increases the level of β-catenin, a downstream substrate of GSK-3β in the Wnt signaling pathway. Shown to selectively induce neuronal differentiation in both pluripotent murine embryonal carcinoma cells (P19; 30-40% at 1 µM) and embryonic stem cells (D3; 50-60% at 400 nM).

Biochem/physiol Actions

Cell permeable: yes
Kd = 126 nM against GSK-3β
Primary Target
GSK-3β
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Carcinogenic / Teratogenic (D)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Ding, S., et al. 2003. Proc. Natl. Acad. Sci. USA100, 7632.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ho Ngai et al.
Cancer immunology research, 11(2), 171-183 (2022-12-10)
Vα24-invariant natural killer T cells (NKT) possess innate antitumor properties that can be exploited for cancer immunotherapy. We have shown previously that the CD62L+ central memory-like subset of these cells drives the in vivo antitumor activity of NKTs, but molecular

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service