Skip to Content
Merck
All Photos(2)

Documents

36-010

Sigma-Aldrich

Anti-phospho-MKK7/SKK4 (Thr275/Ser277) Antibody

Upstate®, from rabbit

Synonym(s):

Dual specificity mitogen-activated protein kinase kinase 7, EC:2.7.12.2, MAP kinase kinase 7, MAPKK 7, JNK-activating kinase 2, MAPK/ERK kinase 7, MEK 7, Stress-activated protein kinase kinase 4, SKK4, SAPK kinase 4, SAPKK-4, SAPKK4, c-Jun N-terminal kin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

rat (based on 100% sequence homology), mouse (based on 100% sequence homology)

manufacturer/tradename

Upstate®

technique(s)

western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

phosphorylation (pThr275/pSer277)

Gene Information

human ... MAP2K7(5609)

General description

Dual specificity mitogen-activated protein kinase kinase 7 (UniProt: O14733; also known as EC:2.7.12.2, MAP kinase kinase 7, MAPKK 7, JNK-activating kinase 2, MAPK/ERK kinase 7, MEK 7;Stress-activated protein kinase kinase 4, SKK4, SAPK kinase 4, SAPKK-4, SAPKK4, c-Jun N-terminal kinase kinase 2,JNK kinase 2, JNKK 2) is encoded by the MAP2K7 (also known as JNKK2, MEK7, MKK7, RKMK7, SKK4) gene (Gene ID: 5609) in human. MKK4/SKK4 belongs to the STE Ser/Thr protein kinase family and is an essential component of the MAP kinase signal transduction pathway. MKK4/SKK4 is ubiquitously expressed with highest level of expression reported in skeletal muscle. It contains a single protein kinase domain and has a DVD domain (aa 377-400) that contains a conserved docking site and bind to their specific upstream MAP kinase kinase kinases (MAPK3Ks). MKK4/SKK4 also has a D domain in the N-terminal region (aa 37-57), which contains a conserved docking site that is required for the binding of MAPK substrates. It is activated by phosphorylation on Ser271 and Thr275 by MAP kinase kinase kinases (MAP3Ks) and it directly phosphorylates and activates the c-Jun NH2-terminal kinases (JNK), in response to cellular stresses and pro-inflammatory cytokines. Mice lacking the MKK4 gene are shown to have abnormal hepatogenesis and die before embryonic day 14.

Specificity

This rabbit polyclonal antibody detects MKK7/SKK4 phosphorylated on Thr275/Ser277.

Immunogen

peptide corresponding to amino acids surrounding Thr275/Ser277 of human MKK7/SKK4.

Application

Anti-Phospho-MKK7/SKK4 (Thr275/Ser277), Cat. No. 36-010, is a highly specific rabbit polyclonal antibody that detect dual specificity mitogen-activated protein kinase kinase 7 (MKK7) and has been tested for use in Western Blotting.

Quality

Evaluated by Western Blotting in lysate of A431 cells treates with Calyculin A and okadaic acid.

Western Blotting Analysis: A 1:500 dilution of this antibody detected Phospho-MKK7/SKK4 (Thr275/Ser277) in lysate from 10 µg of A431 cells treated with Calyculin A and okadaic acid.

Target description

~47kDa

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Yoshida et al.
Molecular and cellular biology, 20(15), 5370-5380 (2000-07-13)
The cellular response to DNA damage includes activation of the nuclear Lyn protein tyrosine kinase. Using cells deficient in Lyn expression, the present studies demonstrate that Lyn is required in part for induction of the stress-activated protein kinase (SAPK) in
I N Foltz et al.
The Journal of biological chemistry, 273(15), 9344-9351 (1998-05-16)
We report the cloning of a novel human activator of c-Jun N-terminal kinase (JNK), mitogen-activated protein kinase kinase 7 (MKK7). The mRNA for MKK7 is widely expressed in humans and mice and encodes a 47-kDa protein (419 amino acids), as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service