346351
D-(+)-Glucose
anhydrous
Synonym(s):
Dextrose, α-D-Glucose, Dextrose
About This Item
Recommended Products
grade
anhydrous
Quality Level
form
powder
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
impurities
≤0.2% Maltose
color
white
mp
150-152 °C (lit.)
solubility
water: soluble
cation traces
heavy metals: ≤0.001%
shipped in
ambient
storage temp.
10-30°C
SMILES string
OC[C@H]1O[C@H](O)[C@H](O)[C@@H](O)[C@@H]1O
InChI
1S/C6H12O6/c7-1-2-3(8)4(9)5(10)6(11)12-2/h2-11H,1H2/t2-,3-,4+,5-,6+/m1/s1
InChI key
WQZGKKKJIJFFOK-DVKNGEFBSA-N
Looking for similar products? Visit Product Comparison Guide
Warning
Reconstitution
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service