Skip to Content
Merck
All Photos(1)

Key Documents

345810

Millipore

G 418 Sulfate, Cell Culture Tested

G418 also known as Geneticin is an aminoglycoside antibiotic related to Gentamicin. Used as a selective agent in transfection of eukaryotic cells. Has highest potency ≥730 µg/mg and purity ≥98%.

Synonym(s):

G 418 Sulfate, Cell Culture Tested, G418, G 418, NMD Inhibitor I

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H40N4O10 · 2H2SO4
CAS Number:
Molecular Weight:
692.71
MDL number:
UNSPSC Code:
12352200
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (TLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

impurities

≤1.0% Ammonia

color

white

solubility

water: 100 mg/mL, clear, colorless

cation traces

heavy metals: ≤100 ppm

shipped in

ambient

storage temp.

10-30°C

InChI

1S/C20H40N4O10.2H2O4S/c1-6(25)14-11(27)10(26)9(23)18(32-14)33-15-7(21)4-8(22)16(12(15)28)34-19-13(29)17(24-3)20(2,30)5-31-19;2*1-5(2,3)4/h6-19,24-30H,4-5,21-23H2,1-3H3;2*(H2,1,2,3,4)/t6?,7-,8+,9+,10+,11-,12-,13-,14+,15+,16-,17-,18+,19-,20+;;/m0../s1

InChI key

UHEPSJJJMTWUCP-NKCAIAFTSA-N

General description

Also available as a sterile-filtered aqueous solution (Cat. No. 345812), a sterile filtered solution in 100 mM HEPES (Cat. No. 508940), and a sterile-filtered solution PBS (Cat. No. 509227).

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, sterilize by filtration through a 0.22 µm or 0.45 µm pore size filter, aliquot and freeze (-20°C) for long term storage or refrigerate (4°C) for short-term storage. Sterile stock solutions are stable for up to 1 year at -20°C.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qin Zhou et al.
Biotechnology journal, 16(9), e2000623-e2000623 (2021-05-31)
Chinese hamster ovary (CHO) cells with a high viable cell density (VCD), resilience to culture stress, and the capacity to continuously express recombinant proteins are highly desirable. Phosphatase and tension homology deleted on chromosome ten (PTEN) functions as a key
Transgene control engineering in mammalian cells.
Beat P Kramer et al.
Methods in molecular biology (Clifton, N.J.), 308, 123-143 (2005-08-06)
Carlos Barrera-Avalos et al.
iScience, 24(12), 103520-103520 (2021-12-25)
T cell activation requires the processing and presentation of antigenic peptides in the context of a major histocompatibility complex (MHC complex). Cross-dressing is a non-conventional antigen presentation mechanism, involving the transfer of preformed peptide/MHC complexes from whole cells, such as
Wilfried Weber et al.
Methods in molecular biology (Clifton, N.J.), 267, 451-466 (2004-07-23)
Inducible expression of desired transgenes in mammalian cells and animals is a current priority in basic and applied research, biopharmaceutical manufacturing, gene therapy, and tissue engineering, as well as in drug discovery. Among the most prominent human-compatible transgene control technologies
Jiacheng Lin et al.
PLoS pathogens, 18(1), e1010204-e1010204 (2022-01-15)
The hepatitis B virus (HBV) core protein (HBc) functions in multiple steps of the viral life cycle. Heteroaryldihydropyrimidine compounds (HAPs) such as Bay41-4109 are capsid protein allosteric modulators that accelerate HBc degradation and inhibit the virion secretion of HBV, specifically

Articles

Antibiotic kill curve is a dose response experiment in which mammalian cells are subjected to increasing amounts of selection antibiotic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service