Skip to Content
Merck
All Photos(1)

Documents

345802

Sigma-Aldrich

Gc-Globulin, Mixed Type, Human Plasma

Native Gc-globulin from human plasma. Single polypeptide chain present in plasma at about 20-55 mg/dl. Functions in binding and transport of vitamin D.

Synonym(s):

Gc-Globulin, Mixed Type, Human Plasma

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352202
NACRES:
NA.25

biological source

human plasma

Quality Level

Assay

≥95% (SDS-PAGE)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

shipped in

ambient

storage temp.

−20°C

General description

Native Gc-globulin from human plasma. A single polypeptide chain present in plasma at a concentration of about 20-55 mg/100 ml. Functions in the binding and transport of vitamin D. May also play an important role in actin homeostasis; binds monomeric actin with high affinity. Binds membranes of both circulating B and T lymphocytes.
Native Gc-globulin from human plasma. Single polypeptide chain present in plasma at about 20-55 mg/dl. Functions in binding and transport of vitamin D.
Native Gc-globulin from human plasma. Single polypeptide chain present in plasma at about 20-55 mg/dl. Functions in binding and transport of vitamin D. May also play an important role in actin homeostasis as it has been shown to bind monomeric actin with high affinity. Binds to cell surface at the DBP binding site in a nonsaturable manner.

Application

Gc-Globulin, Mixed Type, Human Plasma, CAS 68476-36-8, is a single polypeptide chain that transports vitamin D. Binds to cell surface at the DBP binding site in a non-saturable manner.

Warning

Toxicity: Standard Handling (A)

Physical form

Lyophilized from 150 mM NaCl, 20 mM sodium phosphate buffer, pH 7.4.

Preparation Note

Prepared from plasma that has been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV.

Reconstitution

Following reconstitution, aliquot and freeze (-70°C). Stock solutions are stable for up to 1 year at -70°C.

Other Notes

Di Martino, S.J., and Kew, R.R. 1999. J. Immunol. 163, 2135.
Lee, W.M., et al. 1987. Hepatology7, 825.
Machii, T., et al. 1986. Acta Haematol.75, 26.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service