Skip to Content
Merck
All Photos(1)

Key Documents

344931-M

Millipore

Furin Inhibitor II

The Furin Inhibitor II controls the biological activity of Furin. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.

Synonym(s):

Furin Inhibitor II, Hexa-D-arginine, H-(D)RRRRRR-NH₂

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H75N25O6
Molecular Weight:
954.14
UNSPSC Code:
12352200
Pricing and availability is not currently available.

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

white

solubility

water: 1 mg/mL
DMSO: soluble

storage temp.

−20°C

General description

A polyarginine compound that acts as a potent, specific, and competitive inhibitor of furin (Ki = 106 nM). Inhibits Subtilisin-like proprotein convertase 4 (PACE4) and prohormone convertase 1 (PC1) at much higher concentration (Ki = 580 nM and 13 µM, respectively). Has a stimulatory effect on the activity of prohormone convertase 2 (PC2). The D-peptides are suggested to be more resistant to in vivo hydrolysis than L-peptides.
A polyarginine that acts as a potent, specific, and competitive inhibitor of furin (Ki = 106 nM). Inhibits subtilisin-like proprotein convertase 4 (PACE4) and prohormone convertase 1 (PC1) at much higher concentration (Ki = 580 nM and 13 µM, respectively). Does not affect the activity of PC2. The D-peptides are suggested to be more resistant to in vivo hydrolysis than the L-peptides.

Biochem/physiol Actions

Primary Target
furin
Target Ki: 106 nM against Furin

Warning

Toxicity: Standard Handling (A)

Sequence

H-(D)Arg-Arg-Arg-Arg-Arg-Arg-NH₂

Physical form

Supplied as a trifluoroacetate salt.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Cameron, A., et al. 2000. J. Biol. Chem.275, 36741.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service