Skip to Content
Merck
All Photos(1)

Key Documents

344038

Sigma-Aldrich

cFMS Receptor Inhibitor III - CAS 959861-21-3 - Calbiochem

Synonym(s):

cFMS Receptor Inhibitor III - CAS 959861-21-3 - Calbiochem, 4-(3,4-Dimethylanilino)-7-(4-(methylsulfonyl)phenyl)quinoline-3-carboxamide, CSF-1 Receptor Inhibitor III

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H23N3O3S
CAS Number:
Molecular Weight:
445.53
UNSPSC Code:
12352200

Assay

≥97% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 100 mg/mL

shipped in

ambient

storage temp.

2-8°C

General description

This product has been discontinued.



A cell-permeable anilinoquinoline compound that acts as a potent, active site-targeting inhibitor of MCSF receptor/cFMS (IC50 = 8 nM). It is 100-fold more potent in inhibiting the cFMS-dependent growth of M-NSF-60 cells than the cFMS-independent growth of NSO cells (IC50 = 0.1 and 10 µM, respectively).

A cell-permeable anilinoquinoline compound that acts as a potent, active site-targeting inhibitor of MCSF receptor/cFMS (IC50 = 8 nM). It is 100-fold more potent in inhibiting the cFMS-dependent growth of M-NSF-60 cells than the cFMS-independent growth of NSO cells (IC50 = 0.1 and 10 µM, respectively).

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Smalley, T.L. Jr., et al. 2007. Bioorg. Med. Chem. Lett.17, 6257.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service